Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DFFB Full-length ORF (NP_004393.1, 1 a.a. - 338 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001677P01

Catalog No. 89-941-829


Add to cart

Description

Description

Apoptosis is a cell death process that removes toxic and/or useless cells during mammalian development. The apoptotic process is accompanied by shrinkage and fragmentation of the cells and nuclei and degradation of the chromosomal DNA into nucleosomal units. DNA fragmentation factor (DFF) is a heterodimeric protein of 40-kD (DFFB) and 45-kD (DFFA) subunits. DFFA is the substrate for caspase-3 and triggers DNA fragmentation during apoptosis. DFF becomes activated when DFFA is cleaved by caspase-3. The cleaved fragments of DFFA dissociate from DFFB, the active component of DFF. DFFB has been found to trigger both DNA fragmentation and chromatin condensation during apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene but the biological validity of these variants has not been determined. [provided by RefSeq]

Sequence: MLQKPKSVKLRALRSPRKFGVAGRSCQEVLRKGCLRFQLPERGSRLCLYEDGTELTEDYFPSVPDNAELVLLTLGQAWQGYVSDIRRFLSAFHEPQVGLIQAAQQLLCDEQAPQRQRLLADLLHNVSQNIAAETRAEDPPWFEGLESRFQSKSGYLRYSCESRIRSYLREVSSYPSTVGAEAQEEFLRVLGSMCQRLRSMQYNGSYFDRGAKGGSRLCTPEGWFSCQGPFDMDSCLSRHSINPYSNRESRILFSTWNLDHIIEKKRTIIPTLVEAIKEQDGREVDWEYFYGLLFTSENLKLVHIVCHKKTTHKLNCDPSRIYKPQTRLKRKQPVRKRQ
Specifications

Specifications

NP_004393.1
Liquid
1677
DFFB (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLQKPKSVKLRALRSPRKFGVAGRSCQEVLRKGCLRFQLPERGSRLCLYEDGTELTEDYFPSVPDNAELVLLTLGQAWQGYVSDIRRFLSAFHEPQVGLIQAAQQLLCDEQAPQRQRLLADLLHNVSQNIAAETRAEDPPWFEGLESRFQSKSGYLRYSCESRIRSYLREVSSYPSTVGAEAQEEFLRVLGSMCQRLRSMQYNGSYFDRGAKGGSRLCTPEGWFSCQGPFDMDSCLSRHSINPYSNRESRILFSTWNLDHIIEKKRTIIPTLVEAIKEQDGREVDWEYFYGLLFTSENLKLVHIVCHKKTTHKLNCDPSRIYKPQTRLKRKQPVRKRQ
RUO
DFFB
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.5kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAD|CPAN|DFF-40|DFF2|DFF40
DFFB
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit