Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DGCR14 Full-length ORF (AAH37829.1, 1 a.a. - 395 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00008220P01L

 View more versions of this product

Catalog No. 89-941-847


Add to cart

Description

Description

This gene is located within the minimal DGS critical region (MDGCR) thought to contain the gene(s) responsible for a group of developmental disorders. These disorders include DiGeorge syndrome, velocardiofacial syndrome, conotruncal anomaly face syndrome, and some familial or sporadic conotruncal cardiac defects which have been associated with microdeletion of 22q11.2. The encoded protein may be a component of C complex spliceosomes, and the orthologous protein in the mouse localizes to the nucleus. [provided by RefSeq]

Sequence: MLGACFPNLSSGDVMVFGPGGLGQPVLHPVLVLGHHQQPVLGFGSKLVVGPVVGPQACLGVQFALSAVLALPLPLEGGRRRGFGLGGLQPRVAEDLIDVEPLADVGLQHAVDEVLALAGQVLGAREVHTVLLLDTQHLPDVGVVIGHGAADHDVQDHAQAPDVIHLGLVGDALQHLGGCICCRPAEGLAEDDAPIAVPQAALGEAKVRQLDVEVLVEEKVLALEVPVDDVQVVAVLDGGGELSEHLACHVLMQGSLALDELEKRLAPLPSTSPRHVGQQALSSFTLPEASGWAALGWASTQGWDTQHCLRVTSANELIKPRSGGGDWAEGEQGLWNRGSQAGLAGAPTPRPLEGSLSPGQDSGGPAAAALVSLSQVKLEGRRPTSSLFASPGCGD
Specifications

Specifications

AAH37829.1
Liquid
8220
DGCR14 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLGACFPNLSSGDVMVFGPGGLGQPVLHPVLVLGHHQQPVLGFGSKLVVGPVVGPQACLGVQFALSAVLALPLPLEGGRRRGFGLGGLQPRVAEDLIDVEPLADVGLQHAVDEVLALAGQVLGAREVHTVLLLDTQHLPDVGVVIGHGAADHDVQDHAQAPDVIHLGLVGDALQHLGGCICCRPAEGLAEDDAPIAVPQAALGEAKVRQLDVEVLVEEKVLALEVPVDDVQVVAVLDGGGELSEHLACHVLMQGSLALDELEKRLAPLPSTSPRHVGQQALSSFTLPEASGWAALGWASTQGWDTQHCLRVTSANELIKPRSGGGDWAEGEQGLWNRGSQAGLAGAPTPRPLEGSLSPGQDSGGPAAAALVSLSQVKLEGRRPTSSLFASPGCGD
RUO
DGCR14
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
67kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DGCR13|DGS-H|DGS-I|DGSH|DGSI|ES2|Es2el
DGCR14
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit