Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DGCR2 Full-length ORF (NP_005128.1, 1 a.a. - 550 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009993P01S

 View more versions of this product

Catalog No. 89-941-848


Add to cart

Description

Description

Deletions of the 22q11.2 have been associated with a wide range of developmental defects (notably DiGeorge syndrome, velocardiofacial syndrome, conotruncal anomaly face syndrome and isolated conotruncal cardiac defects) classified under the acronym CATCH 22. The DGCR2 gene encodes a novel putative adhesion receptor protein, which could play a role in neural crest cells migration, a process which has been proposed to be altered in DiGeorge syndrome. [provided by RefSeq]

Sequence: MVPKADSGAFLLLFLLVLTVTEPLRPELRCNPGQFACRSGTIQCIPLPWQCDGWATCEDESDEANCPEVTGEVRPHHGKEAVDPRQGRARGGDPSHFHAVNVAQPVRFSSFLGKCPTGWHHYEGTASCYRVYLSGENYWDAAQTCQRLNGSLATFSTDQELRFVLAQEWDQPERSFGWKDQRKLWVGYQYVITGRNRSLEGRWEVAFKGSSEVFLPPDPIFASAMSENDNVFCAQLQCFHFPTLRHHDLHSWHAESCYEKSSFLCKRSQTCVDIKDNVVDEGFYFTPKGDDPCLSCTCHGGEPEMCVAALCERPQGCQQYRKDPKECCKFMCLDPDGNSLFDSMASGMRLVVSCISSFLILSLLLFMVHRLRQRRRERIESLIGANLHHFNLGRRIPGFDYGPDGFGTGLTPLHLSDDGEGGTFHFHDPPPPYTAYKYPDIGQPDDPPPPYEASIHPDSVFYDPADDDAFEPVEVSLPAPGDGGSEGALLRRLEQPLPTAGASLADLEDSADSSSALLVPPDPAQSGSTPAAEALPGGGRHSRSSLNTVV
Specifications

Specifications

NP_005128.1
Liquid
9993
DGCR2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVPKADSGAFLLLFLLVLTVTEPLRPELRCNPGQFACRSGTIQCIPLPWQCDGWATCEDESDEANCPEVTGEVRPHHGKEAVDPRQGRARGGDPSHFHAVNVAQPVRFSSFLGKCPTGWHHYEGTASCYRVYLSGENYWDAAQTCQRLNGSLATFSTDQELRFVLAQEWDQPERSFGWKDQRKLWVGYQYVITGRNRSLEGRWEVAFKGSSEVFLPPDPIFASAMSENDNVFCAQLQCFHFPTLRHHDLHSWHAESCYEKSSFLCKRSQTCVDIKDNVVDEGFYFTPKGDDPCLSCTCHGGEPEMCVAALCERPQGCQQYRKDPKECCKFMCLDPDGNSLFDSMASGMRLVVSCISSFLILSLLLFMVHRLRQRRRERIESLIGANLHHFNLGRRIPGFDYGPDGFGTGLTPLHLSDDGEGGTFHFHDPPPPYTAYKYPDIGQPDDPPPPYEASIHPDSVFYDPADDDAFEPVEVSLPAPGDGGSEGALLRRLEQPLPTAGASLADLEDSADSSSALLVPPDPAQSGSTPAAEALPGGGRHSRSSLNTVV
RUO
DGCR2
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
87.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DGS-C|DKFZp686I1730|IDD|KIAA0163|LAN|SEZ-12
DGCR2
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit