Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DGCR6L Full-length ORF (AAH00682.1, 1 a.a. - 220 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00085359P01L

 View more versions of this product

Catalog No. 89-941-855


Add to cart

Description

Description

This gene, the result of a duplication at this locus, is one of two functional genes encoding nearly identical proteins that have similar expression patterns. The product of this gene is a protein that shares homology with the Drosophila gonadal protein, expressed in gonadal tissues and germ cells, and with the human laminin gamma-1 chain that functions in cell attachment and migration. This gene is located in a region of chromosome 22 implicated in the DiGeorge syndrome, one facet of a broader collection of anomalies referred to as the CATCH 22 syndrome. [provided by RefSeq]

Sequence: MERYAAALEEVADGARQQERHYQLLSALQSLVKELPSSFQQRLSYTTLSDLALALLDGTVFEIVQGLLEIQHLTEKSLYNQRLRLQNEHRVLRQALRQKHQEAQQACRPHNLPVVQAAQQRELEAVEHRIREEQRAMDQKIILELDRKVADQQSTLEKAGVAGFYVTTNPQELMLQMNLLELIRKLQQRGCRAGNAALGLGGPWQSPAAQCDQKGSPVPP
Specifications

Specifications

AAH00682.1
Liquid
85359
DGCR6L (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MERYAAALEEVADGARQQERHYQLLSALQSLVKELPSSFQQRLSYTTLSDLALALLDGTVFEIVQGLLEIQHLTEKSLYNQRLRLQNEHRVLRQALRQKHQEAQQACRPHNLPVVQAAQQRELEAVEHRIREEQRAMDQKIILELDRKVADQQSTLEKAGVAGFYVTTNPQELMLQMNLLELIRKLQQRGCRAGNAALGLGGPWQSPAAQCDQKGSPVPP
RUO
DGCR6L
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
51.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10666
DGCR6L
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit