Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DGKA Full-length ORF (NP_001336.2, 1 a.a. - 735 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001606P02L

 View more versions of this product

Catalog No. 89-941-864


Add to cart

Description

Description

The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It acts as a modulator that competes with protein kinase C for the second messenger diacylglycerol in intracellular signaling pathways. It also plays an important role in the resynthesis of phosphatidylinositols and phosphorylating diacylglycerol to phosphatidic acid. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified. [provided by RefSeq]

Sequence: MAKERGLISPSDFAQLQKYMEYSTKKVSDVLKLFEDGEMAKYVQGDAIGYEGFQQFLKIYLEVDNVPRHLSLALFQSFETGHCLNETNVTKDVVCLNDVSCYFSLLEGGRPEDKLEFTFKLYDTDRNGILDSSEVDKIILQMMRVAEYLDWDVSELRPILQEMMKEIDYDGSGSVSQAEWVRAGATTVPLLVLLGLEMTLKDDGQHMWRPKRFPRPVYCNLCESSIGLGKQGLSCNLCKYTVHDQCAMKALPCEVSTYAKSRKDIGVQSHVWVRGGCESGRCDRCQKKIRIYHSLTGLHCVWCHLEIHDDCLQAVGHECDCGLLRDHILPPSSIYPSVLASGPDRKNSKTSQKTMDDLNLSTSEALRIDPVPNTHPLLVFVNPKSGGKQGQRVLWKFQYILNPRQVFNLLKDGPEIGLRLFKDVPDSRILVCGGDGTVGWILETIDKANLPVLPPVAVLPLGTGNDLARCLRWGGGYEGQNLAKILKDLEMSKVVHMDRWSVEVIPQQTEEKSDPVPFQIINNYFSIGVDASIAHRFHIMREKYPEKFNSRMKNKLWYFEFATSESIFSTCKKLEESLTVEICGKPLDLSNLSLEGIAVLNIPSMHGGSNLWGDTRRPHGDIYGINQALGATAKVITDPDILKTCVPDLSDKRLEVVGLEGAIEMGQIYTKLKNAGRRLAKCSEITFHTTKTLPMQIDGEPWMQTPCTIKITHKNQMPMLMGPPPRSTNFFGFLS
Specifications

Specifications

NP_001336.2
Liquid
1606
DGKA (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAKERGLISPSDFAQLQKYMEYSTKKVSDVLKLFEDGEMAKYVQGDAIGYEGFQQFLKIYLEVDNVPRHLSLALFQSFETGHCLNETNVTKDVVCLNDVSCYFSLLEGGRPEDKLEFTFKLYDTDRNGILDSSEVDKIILQMMRVAEYLDWDVSELRPILQEMMKEIDYDGSGSVSQAEWVRAGATTVPLLVLLGLEMTLKDDGQHMWRPKRFPRPVYCNLCESSIGLGKQGLSCNLCKYTVHDQCAMKALPCEVSTYAKSRKDIGVQSHVWVRGGCESGRCDRCQKKIRIYHSLTGLHCVWCHLEIHDDCLQAVGHECDCGLLRDHILPPSSIYPSVLASGPDRKNSKTSQKTMDDLNLSTSEALRIDPVPNTHPLLVFVNPKSGGKQGQRVLWKFQYILNPRQVFNLLKDGPEIGLRLFKDVPDSRILVCGGDGTVGWILETIDKANLPVLPPVAVLPLGTGNDLARCLRWGGGYEGQNLAKILKDLEMSKVVHMDRWSVEVIPQQTEEKSDPVPFQIINNYFSIGVDASIAHRFHIMREKYPEKFNSRMKNKLWYFEFATSESIFSTCKKLEESLTVEICGKPLDLSNLSLEGIAVLNIPSMHGGSNLWGDTRRPHGDIYGINQALGATAKVITDPDILKTCVPDLSDKRLEVVGLEGAIEMGQIYTKLKNAGRRLAKCSEITFHTTKTLPMQIDGEPWMQTPCTIKITHKNQMPMLMGPPPRSTNFFGFLS
RUO
DGKA
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
109kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DAGK|DAGK1|DGK-alpha|MGC12821|MGC42356
DGKA
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit