Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DGKG Full-length ORF (NP_001074213.1, 1 a.a. - 766 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001608P01S

 View more versions of this product

Catalog No. 89-941-868


Add to cart

Description

Description

This gene encodes an enzyme that is a member of the type I subfamily of diacylglycerol kinases, which are involved in lipid metabolism. These enzymes generate phosphatidic acid by catalyzing the phosphorylation of diacylglycerol, a fundamental lipid second messenger that activates numerous proteins, including protein kinase C isoforms, Ras guanyl nucleotide-releasing proteins and some transient receptor potential channels. Diacylglycerol kinase gamma has been implicated in cell cycle regulation and in the negative regulation of macrophage differentiation in leukemia cells. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MGEERWVSLTPEEFDQLQKYSEYSSKKIKDALTEFNEGGSLKQYDPHEPISYDVFKLFMRAYLEVDLPQPLSTHLFLAFSQKPRHETSDHPTEGASNSEANSADTNIQNADNATKADEACAPDTESNMAEKQAPAEDQVAATPLEPPVPRSSSSESPVVYLKDVVCYLSLLETGRPQDKLEFMFRLYDSDENGLLDQAEMDCIVNQMLHIAQYLEWDPTELRPILKEMLQGMDYDRDGFVSLQEWVHGGMTTIPLLVLLGMDDSGSKGDGRHAWTMKHFKKPTYCNFCHIMLMGVRKQGLCCTYCKYTVHERCVSRNIPGCVKTYSKAKRSGEVMQHAWVEGNSSVKCDRCHKSIKCYQSVTARHCVWCRMTFHRKCELSTLCDGGELRDHILLPTSICPITRDRPGEKSDGCVSAKGELVMQYKIIPTPGTHPLLVLVNPKSGGRQGERLNFFRDTPDFRVLACGGDGTVGWILDCIDKANFAKHPPVAVLPLGTGNDLARCLRWGGGYEGGSLTKILKDIEQSPLVMLDRWHLEVIPREEVENGDQVPYSIMNNYFSIGVDASIAHRFHVMREKHPEKFNSRMKNKLWYFEFGTSETFAATCKKLHDHIELECDGVGVDLSNIFLEGIAILNIPSMYGGTNLWGENKKNRAVIRESRKGVTDPKELKFCVQDLSDQLLEVVGLEGAMEMGQIYTGLKSAGRRLAQCASVTIRTNKLLPMQVDGEPWMQPCCTIKITHKNQAPMMMGPPQKSSFFSLRRKSRSKD
Specifications

Specifications

NP_001074213.1
Liquid
1608
DGKG (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGEERWVSLTPEEFDQLQKYSEYSSKKIKDALTEFNEGGSLKQYDPHEPISYDVFKLFMRAYLEVDLPQPLSTHLFLAFSQKPRHETSDHPTEGASNSEANSADTNIQNADNATKADEACAPDTESNMAEKQAPAEDQVAATPLEPPVPRSSSSESPVVYLKDVVCYLSLLETGRPQDKLEFMFRLYDSDENGLLDQAEMDCIVNQMLHIAQYLEWDPTELRPILKEMLQGMDYDRDGFVSLQEWVHGGMTTIPLLVLLGMDDSGSKGDGRHAWTMKHFKKPTYCNFCHIMLMGVRKQGLCCTYCKYTVHERCVSRNIPGCVKTYSKAKRSGEVMQHAWVEGNSSVKCDRCHKSIKCYQSVTARHCVWCRMTFHRKCELSTLCDGGELRDHILLPTSICPITRDRPGEKSDGCVSAKGELVMQYKIIPTPGTHPLLVLVNPKSGGRQGERLNFFRDTPDFRVLACGGDGTVGWILDCIDKANFAKHPPVAVLPLGTGNDLARCLRWGGGYEGGSLTKILKDIEQSPLVMLDRWHLEVIPREEVENGDQVPYSIMNNYFSIGVDASIAHRFHVMREKHPEKFNSRMKNKLWYFEFGTSETFAATCKKLHDHIELECDGVGVDLSNIFLEGIAILNIPSMYGGTNLWGENKKNRAVIRESRKGVTDPKELKFCVQDLSDQLLEVVGLEGAMEMGQIYTGLKSAGRRLAQCASVTIRTNKLLPMQVDGEPWMQPCCTIKITHKNQAPMMMGPPQKSSFFSLRRKSRSKD
RUO
DGKG
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
112.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DAGK3|DGK-GAMMA|MGC104993|MGC133330
DGKG
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit