Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DGKH Partial ORF (AAH43292, 831 a.a. - 930 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00160851Q01S

 View more versions of this product

Catalog No. 89-941-870


Add to cart

Description

Description

This gene encodes a member of the diacylglycerol kinase (DGK) enzyme family of proteins, specifically the type II DGK subfamily. Members of this family are involved in regulating the intracellular concentrations of diacylglycerol and phosphatidic acid. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: SGKPVLRTHLYIHHAIDLATEEVSQMQLCSQAAEELITRICDAATIHCLLEQELAHAVNACSHALNKANPRCPESLTRDTATEIAINVKALYNETESLLV
Specifications

Specifications

AAH43292
Liquid
160851
DGKH (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DGKeta|DKFZp761I1510
DGKH
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SGKPVLRTHLYIHHAIDLATEEVSQMQLCSQAAEELITRICDAATIHCLLEQELAHAVNACSHALNKANPRCPESLTRDTATEIAINVKALYNETESLLV
RUO
DGKH
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit