Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human DGKZ Full-length ORF (AAH41770.1, 1 a.a. - 929 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00008525P01
Description
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It may attenuate protein kinase C activity by regulating diacylglycerol levels in intracellular signaling cascade and signal transduction. Alternative splicing occurs at this locus and four transcript variants encoding distinct isoforms have been identified. [provided by RefSeq]
Sequence: MEPRDGSPEARSSDSESASASSSGSERDAGPEPDKAPRRLNKRRFPGLRLFGHRKAITKSGLQHLAPPPPTPGAPCSESERQIRSTVDWSESATYGEHIWFETNVSGDFCYVGEQYCVARMLQKSVSRRKCAACKIVVHTPCIEQLEKINFRCKPSFRESGSRNVREPTFVRHHWVHRRRQDGKCRHCGKGFQQKFTFHSKEIVAISCSWCKQAYHSKVSCFMLQQIEEPCSLGVHAAVVIPPTWILRARRPQNTLKASKKKKRASFKRKSSKKGPEEGRWRPFIIRPTPSPLMKPLLVFVNPKSGGNQGAKIIQSFLWYLNPRQVFDLSQGGPKEALEMYRKVHNLRILACGGDGTVGWILSTLDQLRLKPPPPVAILPLGTGNDLARTLNWGGGYTDEPVSKILSHVEEGNVVQLDRWDLHAEPNPEAGPEDRDEGATDRLPLDVFNNYFSLGFDAHVTLEFHESREANPEKFNSRFRNKMFYAGTAFSDFLMGSSKDLAKHIRVVCDGMDLTPKIQDLKPKCVVFLNIPRYCAGTMPWGHPGEHHDFEPQRHDDGYLEVIGFTMTSLAALQVGGHGERLTQCREVVLTTSKAIPVQVDGEPCKLAASRIRIALRNQATMVQKAKRRSAAPLHSDQQPVPEQLRIQVSRVSMHDYEALHYDKEQLKEASVPLGTVVVPGDSDLELCRAHIERLQQEPDGAGAKSPTCQKLSPKWCFLDATTASRFYRIDRAQEHLNYVTEIAQDEIYILDPELLGASARPDLPTPTSPLPTSPCSPTPRSLQGDAAPPQGEELIEAAKRNDFCKLQELHRAGGDLMHRDEQSRTLLHHAVSTGSKDVVRYLLDHAPPEILDAVEENGETCLHQAAALGQRTICHYIVEAGASLMKTDQQGDTPRQRAEKAQDTELAAYLENRQHYQMIQREDQETAVSpecifications
AAH41770.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
130.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DAGK5/DAGK6/DGK-ZETA/hDGKzeta | |
DGKZ | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8525 | |
DGKZ (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEPRDGSPEARSSDSESASASSSGSERDAGPEPDKAPRRLNKRRFPGLRLFGHRKAITKSGLQHLAPPPPTPGAPCSESERQIRSTVDWSESATYGEHIWFETNVSGDFCYVGEQYCVARMLQKSVSRRKCAACKIVVHTPCIEQLEKINFRCKPSFRESGSRNVREPTFVRHHWVHRRRQDGKCRHCGKGFQQKFTFHSKEIVAISCSWCKQAYHSKVSCFMLQQIEEPCSLGVHAAVVIPPTWILRARRPQNTLKASKKKKRASFKRKSSKKGPEEGRWRPFIIRPTPSPLMKPLLVFVNPKSGGNQGAKIIQSFLWYLNPRQVFDLSQGGPKEALEMYRKVHNLRILACGGDGTVGWILSTLDQLRLKPPPPVAILPLGTGNDLARTLNWGGGYTDEPVSKILSHVEEGNVVQLDRWDLHAEPNPEAGPEDRDEGATDRLPLDVFNNYFSLGFDAHVTLEFHESREANPEKFNSRFRNKMFYAGTAFSDFLMGSSKDLAKHIRVVCDGMDLTPKIQDLKPKCVVFLNIPRYCAGTMPWGHPGEHHDFEPQRHDDGYLEVIGFTMTSLAALQVGGHGERLTQCREVVLTTSKAIPVQVDGEPCKLAASRIRIALRNQATMVQKAKRRSAAPLHSDQQPVPEQLRIQVSRVSMHDYEALHYDKEQLKEASVPLGTVVVPGDSDLELCRAHIERLQQEPDGAGAKSPTCQKLSPKWCFLDATTASRFYRIDRAQEHLNYVTEIAQDEIYILDPELLGASARPDLPTPTSPLPTSPCSPTPRSLQGDAAPPQGEELIEAAKRNDFCKLQELHRAGGDLMHRDEQSRTLLHHAVSTGSKDVVRYLLDHAPPEILDAVEENGETCLHQAAALGQRTICHYIVEAGASLMKTDQQGDTPRQRAEKAQDTELAAYLENRQHYQMIQREDQETAV | |
RUO | |
DGKZ | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction