Learn More
Abnova™ Human DHFR (NP_000782.1, 1 a.a. - 187 a.a.) Full-length Recombinant Protein with His tag
Used for Func, SDS-PAGE
Supplier: Abnova™ P3492
Description
Dihydrofolate reductase converts dihydrofolate into tetrahydrofolate, a methyl group shuttle required for the de novo synthesis of purines, thymidylic acid, and certain amino acids. While the functional dihydrofolate reductase gene has been mapped to chromosome 5, multiple intronless processed pseudogenes or dihydrofolate reductase-like genes have been identified on separate chromosomes. Dihydrofolate reductase deficiency has been linked to megaloblastic anemia. [provided by RefSeq]
Sequence: MGSSHHHHHHSSGLVPRGSHMVGSLNCIVAVSQNMGIGKNGDLPWPPLRNEFRYFQRMTTTSSVEGKQNLVIMGKKTWFSIPEKNRPLKGRINLVLSRELKEPPQGAHFLSRSLDDALKLTEQPELANKVDMVWIVGGSSVYKEAMNHPGHLKLFVTRIMQDFESDTFFPEIDLEKYKLLPEYPGVLSDVQEEKGIKYKFEVYEKNDSpecifications
NP_000782.1 | |
Functional Study, SDS-PAGE | |
1719 | |
DHFR (Human) Recombinant Protein | |
Conventional Chromatography | |
100 ug | |
Store at 4°C. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. | |
DHFR | |
Specific activity is 1.5-2.5 units/mL and was obtained by measuring the oxidation of NADPH in absorbance at 340nm during reaction. One unit will convert 1.0μmole of 7,8 dihydrofolate and beta-NADPH to 5,6,7,8-tetrahydrofolate and beta-NADP per minute at | |
Recombinant | |
Escherichia coli expression system | |
>95% by SDS-PAGE |
1 mg/mL | |
Liquid | |
23.6kDa | |
Escherichia coli expression system | |
Loading 3 ug protein in 15% SDS-PAGE | |
MGSSHHHHHHSSGLVPRGSHMVGSLNCIVAVSQNMGIGKNGDLPWPPLRNEFRYFQRMTTTSSVEGKQNLVIMGKKTWFSIPEKNRPLKGRINLVLSRELKEPPQGAHFLSRSLDDALKLTEQPELANKVDMVWIVGGSSVYKEAMNHPGHLKLFVTRIMQDFESDTFFPEIDLEKYKLLPEYPGVLSDVQEEKGIKYKFEVYEKND | |
RUO | |
DHFR | |
E. coli | |
His | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.