Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DHX16 Partial ORF (NP_003578, 940 a.a. - 1041 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$499.00 - $769.00

Specifications

Accession Number NP_003578
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8449
Molecular Weight (g/mol) 36.96kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-941-946
View Documents
Abnova™
H00008449Q01L
25 ug
Each for $769.00
Add to Cart
 
89-941-945
View Documents
Abnova™
H00008449Q01S
10 ug
Each for $499.00
Add to Cart
 
Description

Description

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a functional homolog of fission yeast Prp8 protein involved in cell cycle progression. This gene is mapped to the MHC region on chromosome 6p21.3, a region where many malignant, genetic and autoimmune disease genes are linked. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: RKAITAGYFYHTARLTRSGYRTVKQQQTVFIHPNSSLFEQQPRWLLYHELVLTTKEFMRQVLEIESSWLLEVAPHYYKAKELEDPHAKKMPKKIGKTREELG
Specifications

Specifications

NP_003578
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.96kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DBP2/DDX16/PRO2014/PRP8
DHX16
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
8449
DHX16 (Human) Recombinant Protein (Q01)
RKAITAGYFYHTARLTRSGYRTVKQQQTVFIHPNSSLFEQQPRWLLYHELVLTTKEFMRQVLEIESSWLLEVAPHYYKAKELEDPHAKKMPKKIGKTREELG
RUO
DHX16
Wheat Germ (in vitro)
GST
Liquid
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit