Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DHX32 Full-length ORF (AAH02473.3, 1 a.a. - 743 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00055760P01L

 View more versions of this product

Catalog No. 89-941-952


Add to cart

Description

Description

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a member of this family. The function of this member has not been determined. Alternative splicing of this gene generates 2 transcript variants, but the full length nature of one of the variants has not been defined. [provided by RefSeq]

Sequence: MEEEGLECPNSSSEKRYFPESLDSSDGDEEEVLACEDLELNPFDGLPYSSRYYKLLKEREDLPIWKEKYSFMENLLQNQIVIVSGDAKCGKSAQVPQWCAEYCLSIHYQHGGVICTQVHKQTVVQLALRVADEMDVNIGHEVGYVIPFENCCTNETILRYCTDDMLQREMMSNPFLGSYGVIILDDIHERSIATDVLLGLLKDVLLARPELKLIINSSPHLISKLNSYYGNVPVIEVKNKHPVEVVYLSEAQKDSFESILRLIFEIHHSGEKGDIVVFLACEQDIEKVCETVYQGSNLNPDLGELVVVPLYPKEKCSLFKPLDETEKRCQVYQRRVVLTTSSGEFLIWSNSVRFVIDVGVERRKVYNPRIRANSLVMQPISQSQAEIRKQILGSSSSGKFFCLYTEEFASKDMTPLKPAEMQEANLTSMVLFMKRIDIAGLGHCDFMNRPAPESLMQALEDLDYLAALDNDGNLSEFGIIMSEFPLDPQLSKSILASCEFDCVDEVLTIAAMVTAPNCFSHVPHGAEEAALTCWKTFLHPEGDHFTLISIYKAYQDTTLNSSSEYCVEKWCRDYFLNCSALRMADVIRAELLEIIKRIELPYAEPAFGSKENTLNIKKALLSGYFMQIARDVDGSGNYLMLTHKQVAQLHPLSGYSITKKMPEWVLFHKFSISENNYIRITSEISPELFMQLVPQYYFSNLPPSESKDILQQVVDHLSPVSTMNKEQQMCETCPETEQRCTLQ
Specifications

Specifications

AAH02473.3
Liquid
55760
DHX32 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEEEGLECPNSSSEKRYFPESLDSSDGDEEEVLACEDLELNPFDGLPYSSRYYKLLKEREDLPIWKEKYSFMENLLQNQIVIVSGDAKCGKSAQVPQWCAEYCLSIHYQHGGVICTQVHKQTVVQLALRVADEMDVNIGHEVGYVIPFENCCTNETILRYCTDDMLQREMMSNPFLGSYGVIILDDIHERSIATDVLLGLLKDVLLARPELKLIINSSPHLISKLNSYYGNVPVIEVKNKHPVEVVYLSEAQKDSFESILRLIFEIHHSGEKGDIVVFLACEQDIEKVCETVYQGSNLNPDLGELVVVPLYPKEKCSLFKPLDETEKRCQVYQRRVVLTTSSGEFLIWSNSVRFVIDVGVERRKVYNPRIRANSLVMQPISQSQAEIRKQILGSSSSGKFFCLYTEEFASKDMTPLKPAEMQEANLTSMVLFMKRIDIAGLGHCDFMNRPAPESLMQALEDLDYLAALDNDGNLSEFGIIMSEFPLDPQLSKSILASCEFDCVDEVLTIAAMVTAPNCFSHVPHGAEEAALTCWKTFLHPEGDHFTLISIYKAYQDTTLNSSSEYCVEKWCRDYFLNCSALRMADVIRAELLEIIKRIELPYAEPAFGSKENTLNIKKALLSGYFMQIARDVDGSGNYLMLTHKQVAQLHPLSGYSITKKMPEWVLFHKFSISENNYIRITSEISPELFMQLVPQYYFSNLPPSESKDILQQVVDHLSPVSTMNKEQQMCETCPETEQRCTLQ
RUO
DHX32
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
110.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DDX32|DHLP1|FLJ10694|FLJ10889
DHX32
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit