Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DHX40 Full-length ORF (AAH24187.1, 1 a.a. - 779 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00079665P01

Catalog No. 89-941-955


Add to cart

Description

Description

DDX40 is a member of the DExH/D box family of ATP-dependent RNA helicases. RNA helicases catalyze the unwinding of double-stranded RNA and play a role in RNA metabolism, including pre-mRNA splicing, ribosome biogenesis, and organellar gene expression.[supplied by OMIM]

Sequence: MSRFPAVAGRAPRRQEEGERSRDLQEERLSAVCIADREEKGCTSQEGGTTPTFPIQKQRKKIIQAVRDNSFLIVTGNTGSGKTTQLPKYLYEAGFSQHGMIGVTQPRKVAAISVAQRVAEEMKCTLGSKVGYQVRFDDCSSKETAIKYMTDGCLLKHILGDPNLTKFSVIILDEAHERTLTTDILFGLLKKLFQEKSPNRKEHLKVVVMSATMELAKLSAFFGNCPIFDIPGRLYPVREKFCNLIGPRDRENTAYIQAIVKVTMDIHLNEMAGDILVFLTGQFEIEKSCELLFQMAESVDYDYDVQDTTLDGLLILPCYGSMTTDQQRRIFLPPPPGIRKCVISTNISATSLTIDGIRYVVDGGFVKQLNHNPRLGLDILEVVPISKSEALQRSGRAGRTSSGKCFRIYSKDFWNQCMPDHVIPEIKRTSLTSVVLTLKCLAIHDVIRFPYLDPPNERLILEALKQLYQCDAIDRSGHVTRLGLSMVEFPLPPHLTCAVIKAASLDCEDLLLPIAAMLSVENVFIRPVDPEYQKEAEQRHRELAAKAGGFNDFATLAVIFEQCKSSGAPASWCQKHWIHWRCLFSAFRVEAQLRELIRKLKQQSDFPKETFEGPKHEVLRRCLCAGYFKNVARRSVGRTFCTMDGRGSPVHIHPSSALHEQETKLEWIIFHEVLVTTKVYARIVCPIRYEWVRDLLPKLHEFNAHDLSSVARREVREDARRRWTNKENVKQLKDGISKDVLKKMQRRNDDKSISDARARFLERKQQRTQDHSDTRKETG
Specifications

Specifications

AAH24187.1
Liquid
79665
DHX40 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSRFPAVAGRAPRRQEEGERSRDLQEERLSAVCIADREEKGCTSQEGGTTPTFPIQKQRKKIIQAVRDNSFLIVTGNTGSGKTTQLPKYLYEAGFSQHGMIGVTQPRKVAAISVAQRVAEEMKCTLGSKVGYQVRFDDCSSKETAIKYMTDGCLLKHILGDPNLTKFSVIILDEAHERTLTTDILFGLLKKLFQEKSPNRKEHLKVVVMSATMELAKLSAFFGNCPIFDIPGRLYPVREKFCNLIGPRDRENTAYIQAIVKVTMDIHLNEMAGDILVFLTGQFEIEKSCELLFQMAESVDYDYDVQDTTLDGLLILPCYGSMTTDQQRRIFLPPPPGIRKCVISTNISATSLTIDGIRYVVDGGFVKQLNHNPRLGLDILEVVPISKSEALQRSGRAGRTSSGKCFRIYSKDFWNQCMPDHVIPEIKRTSLTSVVLTLKCLAIHDVIRFPYLDPPNERLILEALKQLYQCDAIDRSGHVTRLGLSMVEFPLPPHLTCAVIKAASLDCEDLLLPIAAMLSVENVFIRPVDPEYQKEAEQRHRELAAKAGGFNDFATLAVIFEQCKSSGAPASWCQKHWIHWRCLFSAFRVEAQLRELIRKLKQQSDFPKETFEGPKHEVLRRCLCAGYFKNVARRSVGRTFCTMDGRGSPVHIHPSSALHEQETKLEWIIFHEVLVTTKVYARIVCPIRYEWVRDLLPKLHEFNAHDLSSVARREVREDARRRWTNKENVKQLKDGISKDVLKKMQRRNDDKSISDARARFLERKQQRTQDHSDTRKETG
RUO
DHX40
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
115kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARG147|DDX40|FLJ22060|FLJ34015|PAD
DHX40
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit