Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DIP2A Partial ORF (NP_055966, 107 a.a. - 301 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00023181Q03L

 View more versions of this product

Catalog No. 89-941-995


Add to cart

Description

Description

The protein encoded by this gene may be involved in axon patterning in the central nervous system. This gene is not highly expressed. Several transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: KYKERKMPMPSKRRSVLVHSSVETYTPPDTSSASEDEGSLRRPGRLTSTPLQSHSSVEPWLDRVIQGSSTSSSASSTSSHPGGRPTTAPSAAATPGAAATTALAGLEAHTHIDLHSAPPDVTTGLVEHSYFERPQVASVRSVPRGCSGSMLETADGVPVNSRVSSKIQQLLNTLKRPKRPPLKEFFVDDFEELLE
Specifications

Specifications

NP_055966
Liquid
23181
DIP2A (Human) Recombinant Protein (Q03)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C21orf106|DIP2
DIP2A
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
47.275kDa
12.5% SDS-PAGE Stained with Coomassie Blue
KYKERKMPMPSKRRSVLVHSSVETYTPPDTSSASEDEGSLRRPGRLTSTPLQSHSSVEPWLDRVIQGSSTSSSASSTSSHPGGRPTTAPSAAATPGAAATTALAGLEAHTHIDLHSAPPDVTTGLVEHSYFERPQVASVRSVPRGCSGSMLETADGVPVNSRVSSKIQQLLNTLKRPKRPPLKEFFVDDFEELLE
RUO
DIP2A
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit