Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DIPA Partial ORF (NP_006839.2, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00011007Q01S

 View more versions of this product

Catalog No. 89-942-000


Add to cart

Description

Description

Hepatitis delta virus (HDV) is a pathogenic human virus whose RNA genome and replication cycle resemble those of plant viroids. Delta-interacting protein A (DIPA), a cellular gene product, has been found to have homology to hepatitis delta virus antigen (HDAg). DIPA interacts with the viral antigen, HDAg, and can affect HDV replication in vitro. [provided by RefSeq]

Sequence: MEAEAGGLEELTDEEMAALGKEELVRRLRREEAARLAALVQRGRLMQEVNRQLQGHLGEIRELKQLNRRLQAENRELRDLCCFLDSERQRGRRAARQWQLFGTQASRAVR
Specifications

Specifications

NP_006839.2
Liquid
11007
DIPA (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DIPA
CCDC85B
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MEAEAGGLEELTDEEMAALGKEELVRRLRREEAARLAALVQRGRLMQEVNRQLQGHLGEIRELKQLNRRLQAENRELRDLCCFLDSERQRGRRAARQWQLFGTQASRAVR
RUO
CCDC85B
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit