Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DISP1 Full-length ORF (AAH07734.1, 1 a.a. - 542 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00084976P01L

 View more versions of this product

Catalog No. 89-942-014


Add to cart

Description

Description

The pattern of cellular proliferation and differentiation that leads to normal development of embryonic structures often depends upon the localized production of secreted protein signals. Cells surrounding the source of a particular signal respond in a graded manner according to the effective concentration of the signal, and this response produces the pattern of cell types constituting the mature structure. A novel segment-polarity gene known as dispatched has been identified in Drosophila and its protein product is required for normal Hedgehog (Hh) signaling. This gene is one of two human homologs of Drosophila dispatched and, based on sequence identity to its mouse counterpart, the encoded protein may play an essential role in Hh patterning activities in the early embryo. [provided by RefSeq]

Sequence: MAMSNGNNDFVVLSNSSIATSAANPSPLTPCDGDHAAQQLTPKEATRTKVSPNGCLQLNGTVKSSFLPLDNQRMPQMLPQCCHPCPYHHPLTSHSSHQECHPEAGPAAPSALASCCMQPHSEYSASLCPNHSPVYQTTCCLQPSPSFCLHHPWPDHFQHQPVQQHIANIRPSRPFKLPKSYAALIADWPVVVLGMCTMFIVVCALVGVLVPELPDFSDPLLGFEPRGTAIGQRLVTWNNMVKNTGYKATLANYPFKYADEQAKSHRDDRWSDDHYEREKREVDWNFHKDSFFCDVPSDRYSRVVFTSSGGETLWNLPAIKSMCNVDNSRIRSHPQFGDLCQRTTAASCCPSWTLGNYIAILNNRSSCQKIVERDVSHTLKLLRTCAKHYQNGTLGPDCWDMAARRKDQLKCTNVPRKCTKYNAVYQILHYLVDKDFMTPKTADYATPALKYSMLFSPTEKGESMMNIYLDNFENWNSSDGVTTITGIEFGIKHSLFQDYLLMDTVYPAIAIVIVLLVMCVYTKSMFITLMTMFAIISSLIVS
Specifications

Specifications

AAH07734.1
Liquid
84976
DISP1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAMSNGNNDFVVLSNSSIATSAANPSPLTPCDGDHAAQQLTPKEATRTKVSPNGCLQLNGTVKSSFLPLDNQRMPQMLPQCCHPCPYHHPLTSHSSHQECHPEAGPAAPSALASCCMQPHSEYSASLCPNHSPVYQTTCCLQPSPSFCLHHPWPDHFQHQPVQQHIANIRPSRPFKLPKSYAALIADWPVVVLGMCTMFIVVCALVGVLVPELPDFSDPLLGFEPRGTAIGQRLVTWNNMVKNTGYKATLANYPFKYADEQAKSHRDDRWSDDHYEREKREVDWNFHKDSFFCDVPSDRYSRVVFTSSGGETLWNLPAIKSMCNVDNSRIRSHPQFGDLCQRTTAASCCPSWTLGNYIAILNNRSSCQKIVERDVSHTLKLLRTCAKHYQNGTLGPDCWDMAARRKDQLKCTNVPRKCTKYNAVYQILHYLVDKDFMTPKTADYATPALKYSMLFSPTEKGESMMNIYLDNFENWNSSDGVTTITGIEFGIKHSLFQDYLLMDTVYPAIAIVIVLLVMCVYTKSMFITLMTMFAIISSLIVS
RUO
DISP1
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
87.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DISPA|DKFZp434I0428|FLJ43740|MGC104180|MGC13130|MGC16796
DISP1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit