Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DKFZp547C195 Partial ORF (NP_997226, 541 a.a. - 640 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00257160Q01L

 View more versions of this product

Catalog No. 89-942-030


Add to cart

Description

Description

Sequence: LAEHERVAASTQPLGRIRALFPAPLAQISTPMFLPSAQVSYPGRSSHAPATCKLCLMCQKLVQPSELHPMACTHVLHKECIKFWAQTNTNDTCPFCPTLK
Specifications

Specifications

NP_997226
Liquid
257160
DKFZp547C195 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp547C195
RNF214
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LAEHERVAASTQPLGRIRALFPAPLAQISTPMFLPSAQVSYPGRSSHAPATCKLCLMCQKLVQPSELHPMACTHVLHKECIKFWAQTNTNDTCPFCPTLK
RUO
RNF214
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit