Learn More
Abnova™ Human DKKL1 Full-length ORF (NP_055234.1, 1 a.a. - 242 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_055234.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 27120 |
Molecular Weight (g/mol) | 53.4kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-942-063
|
Abnova™
H00027120P01L |
25 ug |
Each for $769.00
|
|
|||||
89-942-062
|
Abnova™
H00027120P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
The dickkopf protein family interacts with the Wnt signaling pathway and its members are characterized by two conserved cysteine-rich domains. This gene encodes a secreted protein that has high similarity to the N-terminus of the dickkopf-3 protein and moderate similarity to that protein's C-terminus though the C-terminal cysteine residues are not conserved. [provided by RefSeq]
Sequence: MGEASPPAPARRHLLVLLLLLSTLVIPSAAAPIHDADAQESSLGLTGLQSLLQGFSRLFLKGNLLRGIDSLFSAPMDFRGLPGNYHKEENQEHQLGNNTLSSHLQIDKMTDNKTGEVLISENVVASIQPAEGSFEGDLKVPRMEEKEALVPIQKATDSFHTELHPRVAFWIIKLPRRRSHQDALEGGHWLSEKRHRLQAIRDGLRKGTHKDVLEEGTESSSHSRLSPRKTHLLYILRPSRQLSpecifications
NP_055234.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
53.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGEASPPAPARRHLLVLLLLLSTLVIPSAAAPIHDADAQESSLGLTGLQSLLQGFSRLFLKGNLLRGIDSLFSAPMDFRGLPGNYHKEENQEHQLGNNTLSSHLQIDKMTDNKTGEVLISENVVASIQPAEGSFEGDLKVPRMEEKEALVPIQKATDSFHTELHPRVAFWIIKLPRRRSHQDALEGGHWLSEKRHRLQAIRDGLRKGTHKDVLEEGTESSSHSRLSPRKTHLLYILRPSRQL | |
RUO | |
DKKL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
27120 | |
DKKL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SGY/SGY-1/SGY1 | |
DKKL1 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.