Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DLGAP4 Full-length ORF (NP_892118.1, 1 a.a. - 453 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00022839P01L

 View more versions of this product

Catalog No. 89-942-091


Add to cart

Description

Description

The product of this gene is a membrane-associated guanylate kinase found at the postsynaptic density in neuronal cells. It is a signaling molecule that can interact with potassium channels and receptors, as well as other signaling molecules. The protein encoded by this gene can interact with PSD-95 through its guanylate kinase domain and may be involved in clustering PSD-95 in the postsynaptic density region. The encoded protein is one of at least four similar proteins that have been found. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MALCLELLKQCSSCLVAYKKTPPPVPPRTTSKPFISVTVQSSTESAQDTYLDSQDHKSEVTSQSGLSNSSDSLDSSTRPPSVTRGGVAPAPEAPEPPPKHAALKSEQGTLTSSESHPEAAPKRKLSSIGIQVDCIQPVPKEEPSPATKFQSIGVQVEDDWRSSVPSHSMSSRRDTDSDTQDANDSSCKSSERSLPDCTPHPNSISIDAGPRQAPKIAQIKRNLSYGDNSDPALEASSLPPPDPWLETSSSSPAEPAQPGACRRDGYWFLKLLQAETERLEGWCCQMDKETKENNLSEEVLGKVLSAVGSAQLLMSQKFQQFRGLCEQNLNPDANPRPTAQDLAGFWDLLQLSIEDISMKFDELYHLKANSWQLVETPEKRKEEKKPPPPVPKKPAKSKPAVSRDKASDASDKQRQEARKRLLAAKRAASVRQNSATESADSIEIYVPEAQTRL
Specifications

Specifications

NP_892118.1
Liquid
22839
DLGAP4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MALCLELLKQCSSCLVAYKKTPPPVPPRTTSKPFISVTVQSSTESAQDTYLDSQDHKSEVTSQSGLSNSSDSLDSSTRPPSVTRGGVAPAPEAPEPPPKHAALKSEQGTLTSSESHPEAAPKRKLSSIGIQVDCIQPVPKEEPSPATKFQSIGVQVEDDWRSSVPSHSMSSRRDTDSDTQDANDSSCKSSERSLPDCTPHPNSISIDAGPRQAPKIAQIKRNLSYGDNSDPALEASSLPPPDPWLETSSSSPAEPAQPGACRRDGYWFLKLLQAETERLEGWCCQMDKETKENNLSEEVLGKVLSAVGSAQLLMSQKFQQFRGLCEQNLNPDANPRPTAQDLAGFWDLLQLSIEDISMKFDELYHLKANSWQLVETPEKRKEEKKPPPPVPKKPAKSKPAVSRDKASDASDKQRQEARKRLLAAKRAASVRQNSATESADSIEIYVPEAQTRL
RUO
DLGAP4
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
75.90kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DAP4|KIAA0964|MGC131862|RP5-977B1.6|SAPAP4
DLGAP4
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit