Learn More
Human DLL4 Fc Chimera, Recombinant Protein, Invitrogen™
HEK293 cells Recombinant Protein
Manufacturer:  Thermo Scientific RP87690
Encompass
 View more versions of this product
Catalog No. PIRP87690
Add to cart
Description
Description
Severe Acute Respiratory Syndrome (SARS), an emerging disease characterized by atypical pneumonia, has recently been attributed to a novel coronavirus (SARS-CoV). The nucleocapsid protein (N) was one of the structural proteins of SARS-CoV (N, S, sE, M). The severe acute respiratory syndrome coronavirus (SARS-CoV) nucleocapsid (N) protein is the most abundant protein in the virus-infected cells. The SARS-CoV-2 S gene has 76% amino acid similarity to the SARS-CoV S gene, and nonsynonymous mutations developed in the S protein as the SARS-CoV epidemic developed, reports have shown that the crystal structure of the SARS-CoV-2 nucleocapsid protein is similar to those of previously described coronavirus N proteins, but their surface electrostatic potential characteristics are distinct.
Specifications
Specifications
Control, ELISA, Western Blot
PBS with no preservative
>95%
HEK293 cells
RUO
Virus
Yes
Lyophilized
SARS-CoV-2 Nucleocapsid (aa1-419), His Tag
1 mg
-20°Cor to +80°C if preferred
Coronavirus; COVID; COVID 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Nucleocapsid; SARS-CoV2; SARS-CoV-2
Unlabeled