Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DLL4 Fc Chimera, Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

HEK293 cells Recombinant Protein

$3430.00

Specifications

Recombinant Yes
Name SARS-CoV-2 Nucleocapsid (aa1-419), His Tag
For Use With (Application) Control, ELISA, Western Blot
Conjugate Unlabeled
Format Lyophilized
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87690 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87690
1 mg
Each for $3,430.00
Add to cart
 
Description

Description

Delta-like ligand 4 (DLL4), also know as Delta-4, is a homologue of the Drosophila delta protein, and functions as a transmembrane bound ligand to the notch receptor Notch1. DLL4 is expressed in the vasculature and plays a critical role in vascular development. It is induced by vascular endothelial growth factor (VEGF), as a negative feedback regulator to regulate angiogenic sprouting and promote the formulation of a differentiated vascular network. DLL4 has been found to be strongly expressed in tumor vessels of clear-cell renal tumors and bladder cancer, and inhibition of DLL4 results in increased vascular proliferation but defective maturation. This in turn leads to a decrease in tumor growth, with no apparent toxicity.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Lyophilized
Coronavirus; COVID; COVID 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Nucleocapsid; SARS-CoV2; SARS-CoV-2
RUO
Virus
SARS-CoV-2 Nucleocapsid (aa1-419), His Tag
Unlabeled
PBS with no preservative
HEK293 cells
-20°Cor to +80°C if preferred
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit