Learn More
Abnova™ Human DMC1 Partial ORF (NP_008999, 237 a.a. - 339 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00011144Q01L
Description
The protein encoded by this gene is essential for meiotic homologous recombination. Genetic recombination in meiosis plays an important role in generating diversity of genetic information. The product of this gene is structurally and evolutionary related to the products of the yeast RAD51 and E. coli RecA genes. Alternative splice variants of this gene have been described but their full-length nature has not been determined. [provided by RefSeq]
Sequence: GELAERQQKLAQMLSRLQKISEEYNVAVFVTNQMTADPGATMTFQADPKKPIGGHILAHASTTRISLRKGRGELRIAKIYDSPEMPENEATFAITAGGIGDAKSpecifications
NP_008999 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.07kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GELAERQQKLAQMLSRLQKISEEYNVAVFVTNQMTADPGATMTFQADPKKPIGGHILAHASTTRISLRKGRGELRIAKIYDSPEMPENEATFAITAGGIGDAK | |
RUO | |
DMC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11144 | |
DMC1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DMC1H/HsLim15/LIM15/MGC150472/MGC150473/dJ199H16.1 | |
DMC1 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.