Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DMGDH Partial ORF (NP_037523.2, 556 a.a. - 665 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00029958Q01S

 View more versions of this product

Catalog No. 89-942-132


Add to cart

Description

Description

This gene encodes an enzyme involved in the catabolism of choline, catalyzing the oxidative demethylation of dimethylglycine to form sarcosine. The enzyme is found as a monomer in the mitochondrial matrix, and uses flavin adenine dinucleotide and folate as cofactors. Mutation in this gene causes dimethylglycine dehydrogenase deficiency, characterized by a fishlike body odor, chronic muscle fatigue, and elevated levels of the muscle form of creatine kinase in serum. [provided by RefSeq]

Sequence: ANVIPKVGFTNISHMLTPKGRVYAELTVSHQSPGEFLLITGSGSELHDLRWIEEEAVKGGYDVEIKNITDELGVLGVAGPQARKVLQKLTSEDLSDDVFKFLQTKSLKVS
Specifications

Specifications

NP_037523.2
Liquid
29958
DMGDH (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DMGDHD|ME2GLYDH
DMGDH
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ANVIPKVGFTNISHMLTPKGRVYAELTVSHQSPGEFLLITGSGSELHDLRWIEEEAVKGGYDVEIKNITDELGVLGVAGPQARKVLQKLTSEDLSDDVFKFLQTKSLKVS
RUO
DMGDH
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit