Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DNAJB12 Full-length ORF (NP_001002762.1, 1 a.a. - 375 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00054788P01

Catalog No. 89-942-170


Add to cart

Description

Description

DNAJB12 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 [PubMed 11147971]).[supplied by OMIM]

Sequence: MESNKDEAERCISIALKAIQSNQPDRALRFLEKAQRLYPTPRVRALIESLNQKPQTAGDQPPPTDTTHATHRKAGGTDAPSANGEAGGESTKGYTAEQVAAVKRVKQCKDYYEILGVSRGASDEDLKKAYRRLALKFHPDKNHAPGATEAFKAIGTAYAVLSNPEKRKQYDQFGDDKSQAARHGHGHGDFHRGFEADISPEDLFNMFFGGGFPSSNVHVYSNGRMRYTYQQRQDRRDNQGDGGLGVFVQLMPILILILVSALSQLMVSSPPYSLSPRPSVGHIHRRVTDHLGVVYYVGDTFSEEYTGSSLKTVERNVEDDYIANLRNNCWKEKQQKEGLLYRARYFGDTDMYHRAQKMGTPSCSRLSEVQASLHG
Specifications

Specifications

NP_001002762.1
Liquid
54788
DNAJB12 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MESNKDEAERCISIALKAIQSNQPDRALRFLEKAQRLYPTPRVRALIESLNQKPQTAGDQPPPTDTTHATHRKAGGTDAPSANGEAGGESTKGYTAEQVAAVKRVKQCKDYYEILGVSRGASDEDLKKAYRRLALKFHPDKNHAPGATEAFKAIGTAYAVLSNPEKRKQYDQFGDDKSQAARHGHGHGDFHRGFEADISPEDLFNMFFGGGFPSSNVHVYSNGRMRYTYQQRQDRRDNQGDGGLGVFVQLMPILILILVSALSQLMVSSPPYSLSPRPSVGHIHRRVTDHLGVVYYVGDTFSEEYTGSSLKTVERNVEDDYIANLRNNCWKEKQQKEGLLYRARYFGDTDMYHRAQKMGTPSCSRLSEVQASLHG
RUO
DNAJB12
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
68.3kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DJ10|DKFZp586B2023
DNAJB12
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit