Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DNAJB7 Full-length ORF (NP_660157.1, 1 a.a. - 309 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00150353P01L

 View more versions of this product

Catalog No. 89-942-187


Add to cart

Description

Description

DNAJB7 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 [PubMed 11147971]).[supplied by OMIM]

Sequence: MVDYYEVLGLQRYASPEDIKKAYHKVALKWHPDKNPENKEEAERKFKEVAEAYEVLSNDEKRDIYDKYGTEGLNGGGSHFDDECEYGFTFHKPDDVFKEIFHERDPFSFHFFEDSLEDLLNRPGSSYGNRNRDAGYFFSTASEYPIFEKFSSYDTGYTSQGSLGHEGLTSFSSLAFDNSGMDNYISVTTSDKIVNGRNINTKKIIESDQEREAEDNGELTFFLVNSVANEEGFAKECSWRTQSFNNYSPNSHSSKHVSQYTFVDNDEGGISWVTSNRDPPIFSAGVKEGGKRKKKKRKEVQKKSTKRNC
Specifications

Specifications

NP_660157.1
Liquid
150353
DNAJB7 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVDYYEVLGLQRYASPEDIKKAYHKVALKWHPDKNPENKEEAERKFKEVAEAYEVLSNDEKRDIYDKYGTEGLNGGGSHFDDECEYGFTFHKPDDVFKEIFHERDPFSFHFFEDSLEDLLNRPGSSYGNRNRDAGYFFSTASEYPIFEKFSSYDTGYTSQGSLGHEGLTSFSSLAFDNSGMDNYISVTTSDKIVNGRNINTKKIIESDQEREAEDNGELTFFLVNSVANEEGFAKECSWRTQSFNNYSPNSHSSKHVSQYTFVDNDEGGISWVTSNRDPPIFSAGVKEGGKRKKKKRKEVQKKSTKRNC
RUO
DNAJB7
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DJ5|HSC3|MGC138340
DNAJB7
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit