Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DNAJB8 Full-length ORF (NP_699161.1, 1 a.a. - 232 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00165721P01S

 View more versions of this product

Catalog No. 89-942-189


Add to cart

Description

Description

DNAJB8 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 [PubMed 11147971]).[supplied by OMIM]

Sequence: MANYYEVLGVQASASPEDIKKAYRKLALRWHPDKNPDNKEEAEKKFKLVSEAYEVLSDSKKRSLYDRAGCDSWRAGGGASTPYHSPFDTGYTFRNPEDIFREFFGGLDPFSFEFWDSPFNSDRGGRGHGLRGAFSAGFGEFPAFMEAFSSFNMLGCSGGSHTTFSSTSFGGSSSGSSGFKSVMSSTEMINGHKVTTKRIVENGQERVEVEEDGQLKSVTVNGKEQLKWMDSK
Specifications

Specifications

NP_699161.1
Liquid
165721
DNAJB8 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MANYYEVLGVQASASPEDIKKAYRKLALRWHPDKNPDNKEEAEKKFKLVSEAYEVLSDSKKRSLYDRAGCDSWRAGGGASTPYHSPFDTGYTFRNPEDIFREFFGGLDPFSFEFWDSPFNSDRGGRGHGLRGAFSAGFGEFPAFMEAFSSFNMLGCSGGSHTTFSSTSFGGSSSGSSGFKSVMSSTEMINGHKVTTKRIVENGQERVEVEEDGQLKSVTVNGKEQLKWMDSK
RUO
DNAJB8
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
52.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DJ6|MGC33884
DNAJB8
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit