Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DNCH1 Partial ORF (AAH21297, 733 a.a. - 832 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001778Q01S

 View more versions of this product

Catalog No. 89-942-254


Add to cart

Description

Description

Dyneins are a group of microtubule-activated ATPases that function as molecular motors. They are divided into two subgroups of axonemal and cytoplasmic dyneins. The cytoplasmic dyneins function in intracellular motility, including retrograde axonal transport, protein sorting, organelle movement, and spindle dynamics. Molecules of conventional cytoplasmic dynein are comprised of 2 heavy chain polypeptides and a number of intermediate and light chains.This gene encodes a member of the cytoplasmic dynein heavy chain family. [provided by RefSeq]

Sequence: TSQGATLDACSFGVTGLKLQGATCNNNKLSLSNAISTALPLTQLRWVKQTNTEKKASVVTLPVYLNFTRADLIFTVDFEIATKEDPRSFYERGVAVLCTE
Specifications

Specifications

AAH21297
Liquid
1778
DNCH1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DHC1|DHC1a|DKFZp686P2245|DNCH1|DNCL|DNECL|DYHC|Dnchc1|HL-3|KIAA0325|p22
DYNC1H1
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TSQGATLDACSFGVTGLKLQGATCNNNKLSLSNAISTALPLTQLRWVKQTNTEKKASVVTLPVYLNFTRADLIFTVDFEIATKEDPRSFYERGVAVLCTE
RUO
DYNC1H1
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit