Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DNMT3L Full-length ORF (AAH02560.1, 1 a.a. - 386 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name DNMT3L (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 69.9kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-942-284
View Documents
Abnova™
H00029947P01L
25 ug
Each for $709.50
Add to cart
 
89-942-283
View Documents
Abnova™
H00029947P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

CpG methylation is an epigenetic modification that is important for embryonic development, imprinting, and X-chromosome inactivation. Studies in mice have demonstrated that DNA methylation is required for mammalian development. This gene encodes a nuclear protein with similarity to DNA methyltransferases. This protein is not thought to function as a DNA methyltransferase as it does not contain the amino acid residues necessary for methyltransferase activity. However, this protein does stimulate de novo methylation by DNA cytosine methyltransferase 3 alpha and it is thought to be required for the establishment of maternal genomic imprints. This protein also mediates transcriptional repression through interaction with histone deacetylase 1. Alternative splicing results in two transcript variants. An additional splice variant has been described but its biological validity has not been determined. [provided by RefSeq]

Sequence: MAAIPALDPEAEPSMDVILVGSSELSSSVSPGTGRDLIAYEVKANQRNIEDICICCGSLQVHTQHPLFEGGICAPCKDKFLDALFLYDDDGYQSYCSICCSGETLLICGNPDCTRCYCFECVDSLVGPGTSGKVHAMSNWVCYLCLPSSRSGLLQRRRKWRSQLKAFYDRESENPLEMFETVPVWRRQPVRVLSLFEDIKKELTSLGFLESGSDPGQLKHVVDVTDTVRKDVEEWGPFDLVYGATPPLGHTCDRPPSWYLFQFHRLLQYARPKPGSPGPFFWMFVDNLVLNKEDLDVASRFLEMEPVTIPDVHGGSLQNAVRVWSNIPAIRSRHWALVSEEELSLLAQNKQSSKLAAKWPTKLVKNCFLPLREYFKYFSTELTSSL
Specifications

Specifications

DNMT3L (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
69.9kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit