Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DOC2B Full-length ORF (AAI53025.1, 1 a.a. - 412 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00008447P01L

 View more versions of this product

Catalog No. 89-942-295


Add to cart

Description

Description

There are at least two protein isoforms of the Double C2 protein, namely alpha (DOC2A) and beta (DOC2B), which contain two C2-like domains. DOC2A and DOC2B are encoded by different genes; these genes are at times confused with the unrelated DAB2 gene which was initially named DOC-2. DOC2B is expressed ubiquitously and is suggested to be involved in Ca(2+)-dependent intracellular vesicle trafficking in various types of cells. [provided by RefSeq]

Sequence: MTLRRRGEKATISIQEHMAIDVCPGPIRPIKQISDYFPRFPRGLPPDAGPRAAAPPDAPARPAVAGAGRRSPSDGAREDDEDVDQLFGAYGSSPGPSPGPSPARPPAKPPEDEPDADGYESDDCTALGTLDFSLLYDQENNALHCTITKAKGLKPMDHNGLADPYVKLHLLPGASKANKLRTKTLRNTLNPTWNETLTYYGITDEDMIRKTLRISVCDEDKFRHNEFIGETRVPLKKLKPNHTKTFNICLEKQLPVDKTEDKSLEERGRILISLKYSSQKQGLLVGIVRCAHLAAMDANGYSDPYVKTYLRPDVDKKSKHKTAVKKKTLNPEFNEEFCYEIKHGDLAKKSLEVTVWDYDIGKSNDFIGGVVLGIHAKGERLKHWFDCLKNKDKRIERWHTLTSELPGAVLSD
Specifications

Specifications

AAI53025.1
Liquid
8447
DOC2B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTLRRRGEKATISIQEHMAIDVCPGPIRPIKQISDYFPRFPRGLPPDAGPRAAAPPDAPARPAVAGAGRRSPSDGAREDDEDVDQLFGAYGSSPGPSPGPSPARPPAKPPEDEPDADGYESDDCTALGTLDFSLLYDQENNALHCTITKAKGLKPMDHNGLADPYVKLHLLPGASKANKLRTKTLRNTLNPTWNETLTYYGITDEDMIRKTLRISVCDEDKFRHNEFIGETRVPLKKLKPNHTKTFNICLEKQLPVDKTEDKSLEERGRILISLKYSSQKQGLLVGIVRCAHLAAMDANGYSDPYVKTYLRPDVDKKSKHKTAVKKKTLNPEFNEEFCYEIKHGDLAKKSLEVTVWDYDIGKSNDFIGGVVLGIHAKGERLKHWFDCLKNKDKRIERWHTLTSELPGAVLSD
RUO
DOC2B
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
72.27kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DOC2B
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit