Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DOHH Full-length ORF (NP_112594.1, 1 a.a. - 302 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00083475P01S

 View more versions of this product

Catalog No. 89-942-312


Add to cart

Description

Description

This gene encodes a metalloenzyme that catalyzes the last step in the conversion of lysine to the unique amino acid hypusine in eukaryotic initiation factor 5A. The encoded protein hydroxylates deoxyhypusine to form hypusine in the mature eukaryotic initiation factor 5A protein. Alternative splicing results in multiple transcript variants

Sequence: MVTEQEVDAIGQTLVDPKQPLQARFRALFTLRGLGGPGAIAWISQAFDDDSALLKHELAYCLGQMQDARAIPMLVDVLQDTRQEPMVRHEAGEALGAIGDPEVLEILKQYSSDPVIEVAETCQLAVRRLEWLQQHGGEPAAGPYLSVDPAPPAEERDVGRLREALLDESRPLFERYRAMFALRNAGGEEAALALAEGLHCGSALFRHEVGYVLGQLQHEAAVPQLAAALARCTENPMVRHECAEALGAIARPACLAALQAHADDPERVVRESCEVALDMYEHETGRAFQYADGLEQLRGAPS
Specifications

Specifications

NP_112594.1
Liquid
83475
DOHH (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVTEQEVDAIGQTLVDPKQPLQARFRALFTLRGLGGPGAIAWISQAFDDDSALLKHELAYCLGQMQDARAIPMLVDVLQDTRQEPMVRHEAGEALGAIGDPEVLEILKQYSSDPVIEVAETCQLAVRRLEWLQQHGGEPAAGPYLSVDPAPPAEERDVGRLREALLDESRPLFERYRAMFALRNAGGEEAALALAEGLHCGSALFRHEVGYVLGQLQHEAAVPQLAAALARCTENPMVRHECAEALGAIARPACLAALQAHADDPERVVRESCEVALDMYEHETGRAFQYADGLEQLRGAPS
RUO
DOHH
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
59.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HLRC1|MGC4293
DOHH
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit