Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DOK1 Full-length ORF (NP_001372.1, 1 a.a. - 481 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001796P01L

 View more versions of this product

Catalog No. 89-942-315


Add to cart

Description

Description

Docking protein 1 is constitutively tyrosine phosphorylated in hematopoietic progenitors isolated from chronic myelogenous leukemia (CML) patients in the chronic phase. It may be a critical substrate for p210(bcr/abl), a chimeric protein whose presence is associated with CML. Docking protein 1 contains a putative pleckstrin homology domain at the amino terminus and ten PXXP SH3 recognition motifs. Docking protein 2 binds p120 (RasGAP) from CML cells. It has been postulated to play a role in mitogenic signaling. [provided by RefSeq]

Sequence: MDGAVMEGPLFLQSQRFGTKRWRKTWAVLYPASPHGVARLEFFDHKGSSSGGGRGSSRRLDCKVIRLAECVSVAPVTVETPPEPGATAFRLDTAQRSHLLAADAPSSAAWVQTLCRNAFPKGSWTLAPTDNPPKLSALEMLENSLYSPTWEGSQFWVTVQRTEAAERCGLHGSYVLRVEAERLTLLTVGAQSQILEPLLSWPYTLLRRYGRDKVMFSFEAGRRCPSGPGTFTFQTAQGNDIFQAVETAIHRQKAQGKAGQGHDVLRADSHEGEVAEGKLPSPPGPQELLDSPPALYAEPLDSLRIAPCPSQDSLYSDPLDSTSAQAGEGVQRKKPLYWDLYEHAQQQLLKAKLTDPKEDPIYDEPEGLAPVPPQGLYDLPREPKDAWWCQARVKEEGYELPYNPATDDYAVPPPRSTKPLLAPKPQGPAFPEPGTATGSGIKSHNSALYSQVQKSGASGSWDCGLSRVGTDKTGVKSEGST
Specifications

Specifications

NP_001372.1
Liquid
1796
DOK1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDGAVMEGPLFLQSQRFGTKRWRKTWAVLYPASPHGVARLEFFDHKGSSSGGGRGSSRRLDCKVIRLAECVSVAPVTVETPPEPGATAFRLDTAQRSHLLAADAPSSAAWVQTLCRNAFPKGSWTLAPTDNPPKLSALEMLENSLYSPTWEGSQFWVTVQRTEAAERCGLHGSYVLRVEAERLTLLTVGAQSQILEPLLSWPYTLLRRYGRDKVMFSFEAGRRCPSGPGTFTFQTAQGNDIFQAVETAIHRQKAQGKAGQGHDVLRADSHEGEVAEGKLPSPPGPQELLDSPPALYAEPLDSLRIAPCPSQDSLYSDPLDSTSAQAGEGVQRKKPLYWDLYEHAQQQLLKAKLTDPKEDPIYDEPEGLAPVPPQGLYDLPREPKDAWWCQARVKEEGYELPYNPATDDYAVPPPRSTKPLLAPKPQGPAFPEPGTATGSGIKSHNSALYSQVQKSGASGSWDCGLSRVGTDKTGVKSEGST
RUO
DOK1
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
78.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC117395|MGC138860|P62DOK
DOK1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit