Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DOK6 Full-length ORF (NP_689934.2, 1 a.a. - 331 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00220164P01S

 View more versions of this product

Catalog No. 89-942-324


Add to cart

Description

Description

DOK6 is a member of the DOK (see DOK1; MIM 602919) family of intracellular adaptors that play a role in the RET (MIM 164761) signaling cascade (Crowder et al., 2004 [PubMed 15286081]).[supplied by OMIM]

Sequence: MASNFNDIVKQGYVKIRSRKLGIFRRCWLVFKKASSKGPRRLEKFPDEKAAYFRNFHKVTELHNIKNITRLPRETKKHAVAIIFHDETSKTFACESELEAEEWCKHLCMECLGTRLNDISLGEPDLLAAGVQREQNERFNVYLMPTPNLDIYGECTMQITHENIYLWDIHNAKVKLVMWPLSSLRRYGRDSTWFTFESGRMCDTGEGLFTFQTREGEMIYQKVHSATLAIAEQHERLMLEMEQKARLQTSLTEPMTLSKSISLPRSAYWHHITRQNSVGEIYSLQGHGFGSSKMSRAQTFPSYAPEQSEEAQQPLSRSSSYGFSYSSSLIQ
Specifications

Specifications

NP_689934.2
Liquid
220164
DOK6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASNFNDIVKQGYVKIRSRKLGIFRRCWLVFKKASSKGPRRLEKFPDEKAAYFRNFHKVTELHNIKNITRLPRETKKHAVAIIFHDETSKTFACESELEAEEWCKHLCMECLGTRLNDISLGEPDLLAAGVQREQNERFNVYLMPTPNLDIYGECTMQITHENIYLWDIHNAKVKLVMWPLSSLRRYGRDSTWFTFESGRMCDTGEGLFTFQTREGEMIYQKVHSATLAIAEQHERLMLEMEQKARLQTSLTEPMTLSKSISLPRSAYWHHITRQNSVGEIYSLQGHGFGSSKMSRAQTFPSYAPEQSEEAQQPLSRSSSYGFSYSSSLIQ
RUO
DOK6
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DOK5L|HsT3226|MGC20785
DOK6
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit