Learn More
Novus Biologicals™ Human Dopamine D2R/DRD2 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP267264
Description
The Human Dopamine D2R/DRD2 ELISA Kit (Colorimetric) measures human Dopamine D2R/DRD2 in serum, plasma and other biological fluids.Specifications
0.31 - 20 ng/mL | |
This kit recognizes Human D2R/DRD2 in samples. No significant cross-reactivity or interference between Human D2R/DRD2 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
1813 | |
Human | |
GPCR | |
Dopamine D2 R/DRD2 |
0.19 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
DRD2 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'