Learn More
Novus Biologicals™ Human Dopamine D3R/DRD3 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP267267
Catalog No. NBP267267
Add to cart
Description
Description
The Human Dopamine D3R/DRD3 ELISA Kit (Colorimetric) measures human Dopamine D3R/DRD3 in serum, plasma and other biological fluids.
Specifications
Specifications
0.31 - 20 ng/mL
This kit recognizes Human D3R/DRD3 in samples. No significant cross-reactivity or interference between Human D3R/DRD3 and analogues was observed.
1814
Human
1 Kit
Storage is content dependent.
0.19 ng/mL
ELISA
DRD3
ELISA Kit (Colorimetric)
Biological Fluids,Plasma,Serum
Dopamine D3 R/DRD3