Learn More
Novus Biologicals™ Human Dopamine Receptor D4 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP267268
Catalog No. NBP267268
Add to cart
Description
Description
The Human Dopamine Receptor D4 ELISA Kit (Colorimetric) measures human Dopamine Receptor D4 in serum, plasma and other biological fluids.
Specifications
Specifications
0.31 - 20 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
1 Kit
This kit recognizes Human D4R/DRD4 in samples. No significant cross-reactivity or interference between Human D4R/DRD4 and analogues was observed.
1815
0.19 ng/mL
Dopamine Receptor D4
DRD4
Biological Fluids, Plasma, Serum