Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DPF2 Partial ORF (AAH14889, 56 a.a. - 155 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00005977Q01S

 View more versions of this product

Catalog No. 89-942-339


Add to cart

Description

Description

The protein encoded by this gene is a member of the d4 domain family, characterized by a zinc finger-like structural motif. This protein functions as a transcription factor which is necessary for the apoptotic response following deprivation of survival factors. It likely serves a regulatory role in rapid hematopoietic cell growth and turnover. This gene is considered a candidate gene for multiple endocrine neoplasia type I, an inherited cancer syndrome involving multiple parathyroid, enteropancreatic, and pituitary tumors. [provided by RefSeq]

Sequence: WMEKRHRGPGLASGQLYSYPARRWRKKRRAHPPEDPRLSFPSIKPDTDQTLKKEGLISQDGSSLEALLRTDPLEKRGAPDPRVDDDSLGEFPVTNSRARK
Specifications

Specifications

AAH14889
Liquid
5977
DPF2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC10180|REQ|UBID4|ubi-d4
DPF2
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
WMEKRHRGPGLASGQLYSYPARRWRKKRRAHPPEDPRLSFPSIKPDTDQTLKKEGLISQDGSSLEALLRTDPLEKRGAPDPRVDDDSLGEFPVTNSRARK
RUO
DPF2
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit