Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DPH2 Full-length ORF (NP_001375.2, 1 a.a. - 489 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001802P02L

 View more versions of this product

Catalog No. 89-942-344


Add to cart

Description

Description

This gene is one of two human genes similar to the yeast gene dph2. The yeast gene was identified by its ability to complement a diphthamide mutant strain, and thus probably functions in diphthamide biosynthesis. Diphthamide is a post-translationally modified histidine residue present in elongation factor 2 (EF2) that is the target of diphtheria toxin ADP-ribosylation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MESMFSSPAEAALQRETGVPGLLTPLPDLDGVYELERVAGFVRDLGCERVALQFPDQLLGDAVAVAARLEETTGSKMFILGDTAYGSCCVDVLGAEQAGAQALIHFGPACLSPPARPLPVAFVLRQRSVALELCVKAFEAQNPDPKAPVVLLSEPACAHALEALATLLRPRYLDLLVSSPAFPQPVGSLSPEPMPLERFGRRFPLAPGRRLEEYGAFYVGGSKASPDPDLDPDLSRLLLGWAPGQPFSSCCPDTGKTQDEGARAGRLRARRRYLVERARDARVVGLLAGTLGVAQHREALAHLRNLTQAAGKRSYVLALGRPTPAKLANFPEVDVFVLLACPLGALAPQLSGSFFQPILAPCELEAACNPAWPPPGLAPHLTHYADLLPGSPFHVALPPPESELWETPDVSLITGDLRPPPAWKSSNDHGSLALTPRPQLELAESSPAASFLSSRSWQGLEPRLGQTPVTEAVSGRRGIAIAYEDEGSG
Specifications

Specifications

NP_001375.2
Liquid
1802
DPH2 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MESMFSSPAEAALQRETGVPGLLTPLPDLDGVYELERVAGFVRDLGCERVALQFPDQLLGDAVAVAARLEETTGSKMFILGDTAYGSCCVDVLGAEQAGAQALIHFGPACLSPPARPLPVAFVLRQRSVALELCVKAFEAQNPDPKAPVVLLSEPACAHALEALATLLRPRYLDLLVSSPAFPQPVGSLSPEPMPLERFGRRFPLAPGRRLEEYGAFYVGGSKASPDPDLDPDLSRLLLGWAPGQPFSSCCPDTGKTQDEGARAGRLRARRRYLVERARDARVVGLLAGTLGVAQHREALAHLRNLTQAAGKRSYVLALGRPTPAKLANFPEVDVFVLLACPLGALAPQLSGSFFQPILAPCELEAACNPAWPPPGLAPHLTHYADLLPGSPFHVALPPPESELWETPDVSLITGDLRPPPAWKSSNDHGSLALTPRPQLELAESSPAASFLSSRSWQGLEPRLGQTPVTEAVSGRRGIAIAYEDEGSG
RUO
DPH2
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
78.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DPH2L2
DPH2
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit