Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DPM1 Full-length ORF (AAH08466.1, 1 a.a. - 260 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00008813P01

Catalog No. 89-942-357


Add to cart

Description

Description

Dolichol-phosphate mannose (Dol-P-Man) serves as a donor of mannosyl residues on the lumenal side of the endoplasmic reticulum (ER). Lack of Dol-P-Man results in defective surface expression of GPI-anchored proteins. Dol-P-Man is synthesized from GDP-mannose and dolichol-phosphate on the cytosolic side of the ER by the enzyme dolichyl-phosphate mannosyltransferase. Human DPM1 lacks a carboxy-terminal transmembrane domain and signal sequence and is regulated by DPM2. [provided by RefSeq]

Sequence: MASLEVSRGPRRSRRELEVRSPRQNKYSVLLPTYNERENLPLIVWLLVKSFSESGINYEIIIIDDGSPDGTRDVAEQLEKIYGSDRILLRPREKKLGLGTAYIHGMKHATGNYIIIMDADLSHHPKFIPEFIRKQKEGNFDIVSGTRYKGNGGVYGWDLKRKIISRGANFLTQILLRPGASDLTGSFRLYRKEVLEKLIEKCVSKGYVFQMEMIVRARQMNYTIGEVPISFVDRVYGESKLGGNEIVSFLKGLLTLFATT
Specifications

Specifications

AAH08466.1
Liquid
8813
DPM1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASLEVSRGPRRSRRELEVRSPRQNKYSVLLPTYNERENLPLIVWLLVKSFSESGINYEIIIIDDGSPDGTRDVAEQLEKIYGSDRILLRPREKKLGLGTAYIHGMKHATGNYIIIMDADLSHHPKFIPEFIRKQKEGNFDIVSGTRYKGNGGVYGWDLKRKIISRGANFLTQILLRPGASDLTGSFRLYRKEVLEKLIEKCVSKGYVFQMEMIVRARQMNYTIGEVPISFVDRVYGESKLGGNEIVSFLKGLLTLFATT
RUO
DPM1
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
56kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CDGIE|MPDS
DPM1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit