Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DPP10 Full-length ORF (AAH30832.1, 1 a.a. - 796 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00057628P01L

 View more versions of this product

Catalog No. 89-942-363


Add to cart

Description

Description

This gene encodes a single-pass type II membrane protein that is a member of the S9B family in clan SC of the serine proteases. This protein has no detectable protease activity, most likely due to the absence of the conserved serine residue normally present in the catalytic domain of serine proteases. However, it does bind specific voltage-gated potassium channels and alters their expression and biophysical properties. Mutations in this gene have been associated with asthma. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]

Sequence: MNQTASVSHHIKCQPSKTIKELGSNSPPQRNWKGIAIALLVILVVCSLITMSVILLTPDELTNSSETRLSLEDLFRKDFVLHDPEARWINDTDVVYKSENGHVIKLNIETNATTLLLENTTFVTFKASRHSVSPDLKYVLLAYDVKQIFHYSYTASYVIYNIHTREVWELNPPEVEDSVLQYAAWGVQGQQLIYIFENNIYYQPDIKSSSLRLTSSGKEEIIFNGIADWLYEEELLHSHIAHWWSPDGERLAFLMINDSLVPTMVIPRFTGALYPKGKQYPYPKAGQMNPTIKLYVVNLYGPTHTLELMPPDSFKSREYYITMVKWVSNTKTVVRWLNRAQNISILTVCETTTGACSKKYEMTSDTWLSQQNEEPVFSRDGSKFFMTVPVKQGGRGEFHHIAMFLIQSKSEQITVRHLTSGNWEVIKILAYDETTQKIYFLSTESSPRGRQLYSASTEGLLNRQCISCNFMKEQCTYFDASFSPMNQHFLLFCEGPRVPVVSLHSTDNPAKYFILESNSMLKEAILKKKIGKPEIKILHIDDYELPLQLSLPKDFMDRNQYALLLIMDEEPGGQLVTDKFHIDWDSVLIDMDNVIVARFDGRGSGFQGLKILQEIHRRLGSVEVKDQITAVKFLLKLPYIDSKRLSIFGKGYGGYIASMILKSDEKLFKCGSVVAPITDLKLYASAFSERYLGMPSKEESTYQAASVLHNVHGLKEENILIIHGTADTKVHFQHSAELIKHLIKAGVNYTMQVYPDEGHNVSEKSKYHLYSTILKFFSDCLKEEISVLPQEPEEDE
Specifications

Specifications

AAH30832.1
Liquid
57628
DPP10 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNQTASVSHHIKCQPSKTIKELGSNSPPQRNWKGIAIALLVILVVCSLITMSVILLTPDELTNSSETRLSLEDLFRKDFVLHDPEARWINDTDVVYKSENGHVIKLNIETNATTLLLENTTFVTFKASRHSVSPDLKYVLLAYDVKQIFHYSYTASYVIYNIHTREVWELNPPEVEDSVLQYAAWGVQGQQLIYIFENNIYYQPDIKSSSLRLTSSGKEEIIFNGIADWLYEEELLHSHIAHWWSPDGERLAFLMINDSLVPTMVIPRFTGALYPKGKQYPYPKAGQMNPTIKLYVVNLYGPTHTLELMPPDSFKSREYYITMVKWVSNTKTVVRWLNRAQNISILTVCETTTGACSKKYEMTSDTWLSQQNEEPVFSRDGSKFFMTVPVKQGGRGEFHHIAMFLIQSKSEQITVRHLTSGNWEVIKILAYDETTQKIYFLSTESSPRGRQLYSASTEGLLNRQCISCNFMKEQCTYFDASFSPMNQHFLLFCEGPRVPVVSLHSTDNPAKYFILESNSMLKEAILKKKIGKPEIKILHIDDYELPLQLSLPKDFMDRNQYALLLIMDEEPGGQLVTDKFHIDWDSVLIDMDNVIVARFDGRGSGFQGLKILQEIHRRLGSVEVKDQITAVKFLLKLPYIDSKRLSIFGKGYGGYIASMILKSDEKLFKCGSVVAPITDLKLYASAFSERYLGMPSKEESTYQAASVLHNVHGLKEENILIIHGTADTKVHFQHSAELIKHLIKAGVNYTMQVYPDEGHNVSEKSKYHLYSTILKFFSDCLKEEISVLPQEPEEDE
RUO
DPP10
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
117.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DPL2|DPPY|DPRP3
DPP10
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit