Learn More
Abnova™ Human DPP3 Partial ORF (NP_005691.2, 424 a.a. - 504 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010072Q01S
Description
This gene encodes a protein that is a member of the M49 family of metallopeptidases. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH) and has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Increased activity of this protein is associated with endometrial and ovarian cancers. Alternate transcriptional splice variants have been characterized. [provided by RefSeq]
Sequence: LTFLEEDDKDLYILWKGPSFDVQVGLHELLGHGSGKLFVQDEKGAFNFDQETVINPETGEQIQSWYRSGETWDSKFSTIASSpecifications
NP_005691.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.65kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LTFLEEDDKDLYILWKGPSFDVQVGLHELLGHGSGKLFVQDEKGAFNFDQETVINPETGEQIQSWYRSGETWDSKFSTIAS | |
RUO | |
DPP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10072 | |
DPP3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DPPIII/FLJ11387/FLJ22331 | |
DPP3 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.