Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DPP4 (967 amino acids) native Protein from human serum

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P4922

Catalog No. 89-942-374


Add to cart

Description

Description

The protein encoded by this gene is identical to adenosine deaminase complexing protein-2, and to the T-cell activation antigen CD26. It is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. [provided by RefSeq]

Specifications

Specifications

Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
In 20mM Tris-HCl, 500mM NaCl, 1mM CaCl
105kDa
Native protein purified from human serum
>95% by SDS-PAGE
10 ug
RUO
DPP4
Native protein purified from human serum
Human
Liquid
1803
DPP4 (Human) Native Protein
Saltprecipitation/Ion Exchange Chromatography/Gelfiltration
15% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ADABP|ADCP2|CD26|DPPIV|TP103
DPP4
> 200mU/mg
None
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit