Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human DPPIV/CD26 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP191180
Description
The Human DPPIV/CD26 ELISA Kit (Colorimetric) measures human DPPIV/CD26 in cell culture supernatant, serum.Specifications
15.6 - 500ng/mL | |
ELISA Kit | |
The interference of circulating factors of the immune system was evaluated by spiking these proteins at physiologically relevant concentrations into a human DPPIV/CD26 positive serum. No crossreactivity was detected. | |
ELISA | |
DPP4 | |
96 Tests | |
Storage of components varies. See protocol for specific instructions. | |
DPPIV/CD26 |
7.3ng/mL | |
Cell Culture Supernatant, Serum | |
Human | |
1803 | |
Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD26, Biotin-Conjugate anti-human sCD26 monoclonal antibody, Streptavidin-HRP, Human sCD26 Standard, 500ng/mL upon dilution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films | |
Adaptive Immunity, Cellular Markers, Immunology | |
ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, aden |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction