Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DRF1 Full-length ORF (NP_079380.1, 1 a.a. - 170 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00080174P01S

 View more versions of this product

Catalog No. 89-941-463


Add to cart

Description

Description

This gene encodes a regulator of the CDC7-like 1 protein, a serine-threonine kinase which links cell cycle regulation to genome duplication. This protein localizes to the nucleus and its expression is cell cycle-regulated. Alternative splicing of this gene results in two transcript variants encoding different isoforms. Additional transcript variants have been described, but their full length sequences have not been determined. [provided by RefSeq]

Sequence: MSEPGKGDDCLELESSMAESRLRAPDLGVSRCLGKCQKNSPGARKHPFSGKSFYLDLPAGKNLQFLTGAIQQLGGVIEGFLSKEVSYIVSSRREVKAESSGKSHRGCPSPSPSEVRVETSAMVDPKGSHPRPSRKPVDSVPLSRGKELLQKAIRNQVSWGKMGQSRWSPA
Specifications

Specifications

NP_079380.1
Liquid
80174
DRF1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSEPGKGDDCLELESSMAESRLRAPDLGVSRCLGKCQKNSPGARKHPFSGKSFYLDLPAGKNLQFLTGAIQQLGGVIEGFLSKEVSYIVSSRREVKAESSGKSHRGCPSPSPSEVRVETSAMVDPKGSHPRPSRKPVDSVPLSRGKELLQKAIRNQVSWGKMGQSRWSPA
RUO
DBF4B
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ASKL1|DRF1|FLJ13087|MGC15009|ZDBF1B|chifb
DBF4B
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit