Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DRF1 Partial ORF (NP_663696, 201 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00080174Q01L

 View more versions of this product

Catalog No. 89-942-436


Add to cart

Description

Description

This gene encodes a regulator of the CDC7-like 1 protein, a serine-threonine kinase which links cell cycle regulation to genome duplication. This protein localizes to the nucleus and its expression is cell cycle-regulated. Alternative splicing of this gene results in two transcript variants encoding different isoforms. Additional transcript variants have been described, but their full length sequences have not been determined. [provided by RefSeq]

Sequence: VKKQQPKKPEGTCPAAESRTRKVARLKAPFLKIEDESRKFRPFHHQFKSFPEISFLGPKDASPFEAPTTLGSMHHTRESKDGEPSPRSAAHTMPRRKKG
Specifications

Specifications

NP_663696
Liquid
80174
DRF1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ASKL1|DRF1|FLJ13087|MGC15009|ZDBF1B|chifb
DBF4B
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VKKQQPKKPEGTCPAAESRTRKVARLKAPFLKIEDESRKFRPFHHQFKSFPEISFLGPKDASPFEAPTTLGSMHHTRESKDGEPSPRSAAHTMPRRKKG
RUO
DBF4B
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit