Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DSC2 Full-length ORF (NP_004940.1, 1 a.a. - 847 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001824P01L

 View more versions of this product

Catalog No. 89-942-449


Add to cart

Description

Description

The protein encoded by this gene is a calcium-dependent glycoprotein that is a member of the desmocollin subfamily of the cadherin superfamily. These desmosomal family members, along with the desmogleins, are found primarily in epithelial cells where they constitute the adhesive proteins of the desmosome cell-cell junction and are required for cell adhesion and desmosome formation. The desmosomal family members are arranged in two clusters on chromosome 18, occupying less than 650 kb combined. Mutations in this gene are associated with arrhythmogenic right ventricular dysplasia-11. Alternative splicing results in two transcript variants encoding distinct isoforms. [provided by RefSeq]

Sequence: MEAARPSGSWNGALCRLLLLTLAILIFASDACKNVTLHVPSKLDAEKLVGRVNLKECFTAANLIHSSDPDFQILEDGSVYTTNTILLSSEKRSFTILLSNTENQEKKKIFVFLEHQTKVLKKRHTKEKVLRRAKRRWAPIPCSMLENSLGPFPLFLQQVQSDTAQNYTIYYSIRGPGVDQEPRNLFYVERDTGNLYCTRPVDREQYESFEIIAFATTPDGYTPELPLPLIIKIEDENDNYPIFTEETYTFTIFENCRVGTTVGQVCATDKDEPDTMHTRLKYSIIGQVPPSPTLFSMHPTTGVITTTSSQLDRELIDKYQLKIKVQDMDGQYFGLQTTSTCIINIDDVNDHLPTFTRTSYVTSVEENTVDVEILRVTVEDKDLVNTANWRANYTILKGNENGNFKIVTDAKTNEGVLCVVKPLNYEEKQQMILQIGVVNEAPFSREASPRSAMSTATVTVNVEDQDEGPECNPPIQTVRMKENAEVGTTSNGYKAYDPETRSSSGIRYKKLTDPTGWVTIDENTGSIKVFRSLDREAETIKNGIYNITVLASDQGGRTCTGTLGIILQDVNDNSPFIPKKTVIICKPTMSSAEIVAVDPDEPIHGPPFDFSLESSTSEVQRMWRLKAINDTAARLSYQNDPPFGSYVVPITVRDRLGMSSVTSLDVTLCDCITENDCTHRVDPRIGGGGVQLGKWAILAILLGIALLFCILFTLVCGASGTSKQPKVIPDDLAQQNLIVSNTEAPGDDKVYSANGFTTQTVGASAQGVCGTVGSGIKNGGQETIEMVKGGHQTSESCRGAGHHHTLDSCRGGHTEVDNCRYTYSEWHSFTQPRLGEESIRGHTLIKN
Specifications

Specifications

NP_004940.1
Liquid
1824
DSC2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEAARPSGSWNGALCRLLLLTLAILIFASDACKNVTLHVPSKLDAEKLVGRVNLKECFTAANLIHSSDPDFQILEDGSVYTTNTILLSSEKRSFTILLSNTENQEKKKIFVFLEHQTKVLKKRHTKEKVLRRAKRRWAPIPCSMLENSLGPFPLFLQQVQSDTAQNYTIYYSIRGPGVDQEPRNLFYVERDTGNLYCTRPVDREQYESFEIIAFATTPDGYTPELPLPLIIKIEDENDNYPIFTEETYTFTIFENCRVGTTVGQVCATDKDEPDTMHTRLKYSIIGQVPPSPTLFSMHPTTGVITTTSSQLDRELIDKYQLKIKVQDMDGQYFGLQTTSTCIINIDDVNDHLPTFTRTSYVTSVEENTVDVEILRVTVEDKDLVNTANWRANYTILKGNENGNFKIVTDAKTNEGVLCVVKPLNYEEKQQMILQIGVVNEAPFSREASPRSAMSTATVTVNVEDQDEGPECNPPIQTVRMKENAEVGTTSNGYKAYDPETRSSSGIRYKKLTDPTGWVTIDENTGSIKVFRSLDREAETIKNGIYNITVLASDQGGRTCTGTLGIILQDVNDNSPFIPKKTVIICKPTMSSAEIVAVDPDEPIHGPPFDFSLESSTSEVQRMWRLKAINDTAARLSYQNDPPFGSYVVPITVRDRLGMSSVTSLDVTLCDCITENDCTHRVDPRIGGGGVQLGKWAILAILLGIALLFCILFTLVCGASGTSKQPKVIPDDLAQQNLIVSNTEAPGDDKVYSANGFTTQTVGASAQGVCGTVGSGIKNGGQETIEMVKGGHQTSESCRGAGHHHTLDSCRGGHTEVDNCRYTYSEWHSFTQPRLGEESIRGHTLIKN
RUO
DSC2
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
120.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARVD11|CDHF2|DG2|DGII/III|DKFZp686I11137|DSC3
DSC2
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit