Learn More
Abnova™ Human DSC2 Full-length ORF (NP_004940.1, 1 a.a. - 847 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_004940.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1824 |
Molecular Weight (g/mol) | 120.2kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-942-449
|
Abnova™
H00001824P01L |
25 ug |
Each for $769.00
|
|
|||||
89-942-448
|
Abnova™
H00001824P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
The protein encoded by this gene is a calcium-dependent glycoprotein that is a member of the desmocollin subfamily of the cadherin superfamily. These desmosomal family members, along with the desmogleins, are found primarily in epithelial cells where they constitute the adhesive proteins of the desmosome cell-cell junction and are required for cell adhesion and desmosome formation. The desmosomal family members are arranged in two clusters on chromosome 18, occupying less than 650 kb combined. Mutations in this gene are associated with arrhythmogenic right ventricular dysplasia-11. Alternative splicing results in two transcript variants encoding distinct isoforms. [provided by RefSeq]
Sequence: MEAARPSGSWNGALCRLLLLTLAILIFASDACKNVTLHVPSKLDAEKLVGRVNLKECFTAANLIHSSDPDFQILEDGSVYTTNTILLSSEKRSFTILLSNTENQEKKKIFVFLEHQTKVLKKRHTKEKVLRRAKRRWAPIPCSMLENSLGPFPLFLQQVQSDTAQNYTIYYSIRGPGVDQEPRNLFYVERDTGNLYCTRPVDREQYESFEIIAFATTPDGYTPELPLPLIIKIEDENDNYPIFTEETYTFTIFENCRVGTTVGQVCATDKDEPDTMHTRLKYSIIGQVPPSPTLFSMHPTTGVITTTSSQLDRELIDKYQLKIKVQDMDGQYFGLQTTSTCIINIDDVNDHLPTFTRTSYVTSVEENTVDVEILRVTVEDKDLVNTANWRANYTILKGNENGNFKIVTDAKTNEGVLCVVKPLNYEEKQQMILQIGVVNEAPFSREASPRSAMSTATVTVNVEDQDEGPECNPPIQTVRMKENAEVGTTSNGYKAYDPETRSSSGIRYKKLTDPTGWVTIDENTGSIKVFRSLDREAETIKNGIYNITVLASDQGGRTCTGTLGIILQDVNDNSPFIPKKTVIICKPTMSSAEIVAVDPDEPIHGPPFDFSLESSTSEVQRMWRLKAINDTAARLSYQNDPPFGSYVVPITVRDRLGMSSVTSLDVTLCDCITENDCTHRVDPRIGGGGVQLGKWAILAILLGIALLFCILFTLVCGASGTSKQPKVIPDDLAQQNLIVSNTEAPGDDKVYSANGFTTQTVGASAQGVCGTVGSGIKNGGQETIEMVKGGHQTSESCRGAGHHHTLDSCRGGHTEVDNCRYTYSEWHSFTQPRLGEESIRGHTLIKNSpecifications
NP_004940.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
120.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MEAARPSGSWNGALCRLLLLTLAILIFASDACKNVTLHVPSKLDAEKLVGRVNLKECFTAANLIHSSDPDFQILEDGSVYTTNTILLSSEKRSFTILLSNTENQEKKKIFVFLEHQTKVLKKRHTKEKVLRRAKRRWAPIPCSMLENSLGPFPLFLQQVQSDTAQNYTIYYSIRGPGVDQEPRNLFYVERDTGNLYCTRPVDREQYESFEIIAFATTPDGYTPELPLPLIIKIEDENDNYPIFTEETYTFTIFENCRVGTTVGQVCATDKDEPDTMHTRLKYSIIGQVPPSPTLFSMHPTTGVITTTSSQLDRELIDKYQLKIKVQDMDGQYFGLQTTSTCIINIDDVNDHLPTFTRTSYVTSVEENTVDVEILRVTVEDKDLVNTANWRANYTILKGNENGNFKIVTDAKTNEGVLCVVKPLNYEEKQQMILQIGVVNEAPFSREASPRSAMSTATVTVNVEDQDEGPECNPPIQTVRMKENAEVGTTSNGYKAYDPETRSSSGIRYKKLTDPTGWVTIDENTGSIKVFRSLDREAETIKNGIYNITVLASDQGGRTCTGTLGIILQDVNDNSPFIPKKTVIICKPTMSSAEIVAVDPDEPIHGPPFDFSLESSTSEVQRMWRLKAINDTAARLSYQNDPPFGSYVVPITVRDRLGMSSVTSLDVTLCDCITENDCTHRVDPRIGGGGVQLGKWAILAILLGIALLFCILFTLVCGASGTSKQPKVIPDDLAQQNLIVSNTEAPGDDKVYSANGFTTQTVGASAQGVCGTVGSGIKNGGQETIEMVKGGHQTSESCRGAGHHHTLDSCRGGHTEVDNCRYTYSEWHSFTQPRLGEESIRGHTLIKN | |
RUO | |
DSC2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
1824 | |
DSC2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARVD11/CDHF2/DG2/DGII/III/DKFZp686I11137/DSC3 | |
DSC2 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.