Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DSC3 Partial ORF (NP_001932, 585 a.a. - 684 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001825Q01S

 View more versions of this product

Catalog No. 89-942-452


Add to cart

Description

Description

The protein encoded by this gene is a calcium-dependent glycoprotein that is a member of the desmocollin subfamily of the cadherin superfamily. These desmosomal family members, along with the desmogleins, are found primarily in epithelial cells where they constitute the adhesive proteins of the desmosome cell-cell junction and are required for cell adhesion and desmosome formation. The desmosomal family members are arranged in two clusters on chromosome 18, occupying less than 650 kb combined. Alternative splicing results in two transcript variants encoding distinct isoforms. [provided by RefSeq]

Sequence: CKPKMGYTDILAVDPDEPVHGAPFYFSLPNTSPEISRLWSLTKVNDTAARLSYQKNAGFQEYTIPITVKDRAGQAATKLLRVNLCECTHPTQCRATSRST
Specifications

Specifications

NP_001932
Liquid
1825
DSC3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CDHF3|DSC|DSC1|DSC2|DSC4|HT-CP
DSC3
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
CKPKMGYTDILAVDPDEPVHGAPFYFSLPNTSPEISRLWSLTKVNDTAARLSYQKNAGFQEYTIPITVKDRAGQAATKLLRVNLCECTHPTQCRATSRST
RUO
DSC3
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit