Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DSCR1 Partial ORF (AAH02864, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001827Q01L

 View more versions of this product

Catalog No. 89-942-459


Add to cart

Description

Description

The protein encoded by this gene interacts with calcineurin A and inhibits calcineurin-dependent signaling pathways, possibly affecting central nervous system development. This gene is located in the minimal candidate region for the Down syndrome phenotype, and is overexpressed in the brain of Down syndrome fetuses. Chronic overexpression of this gene may lead to neurofibrillary tangles such as those associated with Alzheimer disease. Three transcript variants encoding three different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MEEVDLQDLPSATIACHLDPRVFVDGLCRAKFESLFRTYDKDITFQYFKSFKRVRINFSNPFSAADARLQLHKTEFLGKEMKLYFAQTLHIGSSHLAPPN
Specifications

Specifications

AAH02864
Liquid
1827
DSCR1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ADAPT78|CSP1|DSC1|DSCR1|MCIP1|RCN1
RCAN1
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MEEVDLQDLPSATIACHLDPRVFVDGLCRAKFESLFRTYDKDITFQYFKSFKRVRINFSNPFSAADARLQLHKTEFLGKEMKLYFAQTLHIGSSHLAPPN
RUO
RCAN1
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit