Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DSCR5 Partial ORF (NP_710149, 77 a.a. - 134 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00051227Q01S

 View more versions of this product

Catalog No. 89-942-462


Add to cart

Description

Description

This gene encodes an enzyme involved in the first step of glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid found on many blood cells that serves to anchor proteins to the cell surface. The encoded protein is a component of the GPI-N-acetylglucosaminyltransferase complex that catalyzes the transfer of N-acetylglucosamine (GlcNAc) from UDP-GlcNAc to phosphatidylinositol (PI). This gene is located in the Down Syndrome critical region on chromosome 21 and is a candidate for the pathogenesis of Down syndrome. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]

Sequence: NMMSTSPLDSIHTITDNYAKNQQQKKYQEEAIPALRDISISEVNQMFFLAAKELYTKN
Specifications

Specifications

NP_710149
Liquid
51227
DSCR5 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DCRC|DCRC-S|DSCR5|DSRC
PIGP
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.12kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NMMSTSPLDSIHTITDNYAKNQQQKKYQEEAIPALRDISISEVNQMFFLAAKELYTKN
RUO
PIGP
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit