Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DSE Full-length ORF (NP_037484.1, 1 a.a. - 958 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00029940P01

Catalog No. 89-942-469


Add to cart

Description

Description

The protein encoded by this gene is a tumor-rejection antigen. This antigen possesses tumor epitopes capable of inducing HLA-A24-restricted and tumor-specific cytotoxic T lymphocytes in cancer patients and may be useful for specific immunotherapy. This gene product is localized to the endoplasmic reticulum. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Sequence: MRTHTRGAPSVFFIYLLCFVSAYITDENPEVMIPFTNANYDSHPMLYFSRAEVAELQLRAASSHEHIAARLTEAVHTMLSSPLEYLPPWDPKDYSARWNEIFGNNLGALAMFCVLYPENIEARDMAKDYMERMAAQPSWLVKDAPWDEVPLAHSLVGFATAYDFLYNYLSKTQQEKFLEVIANASGYMYETSYRRGWGFQYLHNHQPTNCMALLTGSLVLMNQGYLQEAYLWTKQVLTIMEKSLVLLREVTDGSLYEGVAYGSYTTRSLFQYMFLVQRHFNINHFGHPWLKQHFAFMYRTILPGFQRTVAIADSNYNWFYGPESQLVFLDKFVMRNGSGNWLADQIRRNRVVEGPGTPSKGQRWCTLHTEFLWYDGSLKSVPPPDFGTPTLHYFEDWGVVTYGSALPAEINRSFLSFKSGKLGGRAIYDIVHRNKYKDWIKGWRNFNAGHEHPDQNSFTFAPNGVPFITEALYGPKYTFFNNVLMFSPAVSKSCFSPWVGQVTEDCSSKWSKYKHDLAASCQGRVVAAEEKNGVVFIRGEGVGAYNPQLNLKNVQRNLILLHPQLLLLVDQIHLGEESPLETAASFFHNVDVPFEETVVDGVHGAFIRQRDGLYKMYWMDDTGYSEKATFASVTYPRGYPYNGTNYVNVTMHLRSPITRAAYLFIGPSIDVQSFTVHGDSQQLDVFIATSKHAYATYLWTGEATGQSAFAQVIADRHKILFDRNSAIKSSIVPEVKDYAAIVEQNLQHFKPVFQLLEKQILSRVRNTASFRKTAERLLRFSDKRQTEEAIDRIFAISQQQQQQSKSKKNRRAGKRYKFVDAVPDIFAQIEVNEKKIRQKAQILAQKELPIDEDEEMKDLLDFADVTYEKHKNGGLIKGRFGQARMVTTTHSRAPSLSASYTRLFLILNIAIFFVMLAMQLTYFQRAQSLHGQRCLYAVLLIDSCILLWLYSSCSQSQC
Specifications

Specifications

NP_037484.1
Liquid
29940
DSE (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRTHTRGAPSVFFIYLLCFVSAYITDENPEVMIPFTNANYDSHPMLYFSRAEVAELQLRAASSHEHIAARLTEAVHTMLSSPLEYLPPWDPKDYSARWNEIFGNNLGALAMFCVLYPENIEARDMAKDYMERMAAQPSWLVKDAPWDEVPLAHSLVGFATAYDFLYNYLSKTQQEKFLEVIANASGYMYETSYRRGWGFQYLHNHQPTNCMALLTGSLVLMNQGYLQEAYLWTKQVLTIMEKSLVLLREVTDGSLYEGVAYGSYTTRSLFQYMFLVQRHFNINHFGHPWLKQHFAFMYRTILPGFQRTVAIADSNYNWFYGPESQLVFLDKFVMRNGSGNWLADQIRRNRVVEGPGTPSKGQRWCTLHTEFLWYDGSLKSVPPPDFGTPTLHYFEDWGVVTYGSALPAEINRSFLSFKSGKLGGRAIYDIVHRNKYKDWIKGWRNFNAGHEHPDQNSFTFAPNGVPFITEALYGPKYTFFNNVLMFSPAVSKSCFSPWVGQVTEDCSSKWSKYKHDLAASCQGRVVAAEEKNGVVFIRGEGVGAYNPQLNLKNVQRNLILLHPQLLLLVDQIHLGEESPLETAASFFHNVDVPFEETVVDGVHGAFIRQRDGLYKMYWMDDTGYSEKATFASVTYPRGYPYNGTNYVNVTMHLRSPITRAAYLFIGPSIDVQSFTVHGDSQQLDVFIATSKHAYATYLWTGEATGQSAFAQVIADRHKILFDRNSAIKSSIVPEVKDYAAIVEQNLQHFKPVFQLLEKQILSRVRNTASFRKTAERLLRFSDKRQTEEAIDRIFAISQQQQQQSKSKKNRRAGKRYKFVDAVPDIFAQIEVNEKKIRQKAQILAQKELPIDEDEEMKDLLDFADVTYEKHKNGGLIKGRFGQARMVTTTHSRAPSLSASYTRLFLILNIAIFFVMLAMQLTYFQRAQSLHGQRCLYAVLLIDSCILLWLYSSCSQSQC
RUO
DSE
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
136.2kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DSEPI|SART2
DSE
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit