Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DSG1 Full-length ORF (AAI53002.1, 1 a.a. - 1049 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001828P01S

 View more versions of this product

Catalog No. 89-942-471

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Desmosomes are cell-cell junctions between epithelial, myocardial and certain other cell types. Desmoglein 1 is a calcium-binding transmembrane glycoprotein component of desmosomes in vertebrate epithelial cells. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. The protein encoded by this gene has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus foliaceus. [provided by RefSeq]

Sequence: MDWSFFRVVAMLFIFLVVVEVNSEFRIQVRDYNTKNGTIKWHSIRRQKREWIKFAAACREGEDNSKRNPIAKIHSDCAANQQVTYRISGVGIDQPPYGIFVINQKTGEINITSIVDREVTPFFIIYCRALNSMGQDLERPLELRVRVLDINDNPPVFSMATFAGQIEENSNANTLVMILNATDADEPNNLNSKIAFKIIRQEPSDSPMFIINRNTGEIRTMNNFLDREQYGQYALAVRGSDRDGGADGMSAECECNIKILDVNDNIPYMEQSSYTIEIQENTLNSNLLEIRVIDLDEEFSANWMAVIFFISGNEGNWFEIEMNERTNVGILKVVKPLDYEAMQSLQLSIGVRNKAEFHHSIMSQYKLKASAISVTVLNVIEGPVFRPGSKTYVVTGNMGSNDKVGDFVATDLDTGRPSTTVRYVMGNNPADLLAVDSRTGKLTLKNKVTKEQYNMLGGKYQGTILSIDDNLQRTCTGTININIQSFGNDDRTNTEPNTKITTNTGRQESTSSTNYDTSTTSTDSSQVYSSEPGNGAKDLLSDNVHFGPAGIGLLIMGFLVLGLVPFLMICCDCGGAPRSAAGFEPVPECSDGAIHSWAVEGPQPEPRDITTVIPQIPPDNANIIECIDNSGVYTNEYGGREMQDLGGGERMTGFELTEGVKTSGMPEICQEYSGTLRRNSMRECREGGLNMNFMESYFCQKAYAYADEDEGRPSNDCLLIYDIEGVGSPAGSVGCCSFIGEDLDDSFLDTLGPKFKKLADISLGKESYPDLDPSWPPQSTEPVCLPQETEPVVSGHPPISPHFGTTTVISESTYPSGPGVLHPKPILDPLGYGNVTVTESYTTSDTLKPSVHVHDNRPASNVVVTERVVGPISGADLHGMLEMPDLRDGSNVIVTERVIAPSSSLPTSLTIHHPRESSNVVVTERVIQPTSGMIGSLSMHPELANAHNVIVTERVVSGAGVTGISGTTGISGGIGSSGLVGTSMGAGSGALSGAGISGGGIGLSSLGGTASIGHMRSSSDHHFNQTIGSASPSTARSRITKYSTVQYSK
Specifications

Specifications

AAI53002.1
Liquid
1828
Human DSG1 Full-length ORF (AAI53002.1, 1 a.a. - 1049 a.a.) Recombinant Protein with GST-tag at N-terminal
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at −80°C. Aliquot to avoid repeated freezing and thawing.
CDHF4, DG1, DSG
DSG1
Wheat Germ (in vitro)
GST Tag
AP, Array, ELISA, WB-Re
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
142.34kDa
Glutathione Sepharose 4 Fast Flow
10μg
RUO
DSG1
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit